Skip to Content
Merck
All Photos(1)

Key Documents

E9904

Sigma-Aldrich

Ergtoxin Peptide

>98% (HPLC), lyophilized powder, recombinant, expressed in E. coli

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

Product Name

Ergtoxin from Centruroides noxius (Scorpion), recombinant, expressed in E. coli, >98% (HPLC), lyophilized powder

recombinant

expressed in E. coli

Quality Level

Assay

>98% (HPLC)

form

lyophilized powder

mol wt

apparent mol wt ~4.7 kDa by SDS-PAGE

solubility

aqueous buffer: soluble

storage temp.

−20°C

SMILES string

N%16%15[C@@](O[C@](C%16=O)(NC(=O)[C@H]%19CN([C@@H]%20Cc%21c%22c([nH]c%21)cccc%22C%20=C%19)C)C(C)C)([C@H]%17N(CCC%17)C(=O)[C@@H]%15Cc%18ccccc%18)O.N98[C@@](O[C@](C9=O)(NC(=O)[C@H]%11CN([C@@H]%12Cc%13c%14c([nH]c%13)cccc%14C%12=C%11)C)C(C)C)([C@H]%10N(CCC%10

InChI

1S/C35H39N5O5.C32H41N5O5.C31H39N5O5/c1-20(2)34(37-31(41)23-16-25-24-11-7-12-26-30(24)22(18-36-26)17-27(25)38(3)19-23)33(43)40-28(15-21-9-5-4-6-10-21)32(42)39-14-8-13-29(39)35(40,44)45-34;1-17(2)12-25-29(39)36-11-7-10-26(36)32(41)37(25)30(40)31(42-32,18(3)

InChI key

QOTUIZVGUVLMNQ-SBJOBWOOSA-N

Biochem/physiol Actions

A novel ERG1 (KV11.1) K+ channel blocker

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miguel Corona et al.
FEBS letters, 532(1-2), 121-126 (2002-12-03)
Twenty-three novel sequences similar to Ergtoxin (ErgTx) were obtained by direct sequencing of peptides or deduced from gene cloned using cDNAs of venomous glands of Centruroides (C.) elegans, C. exilicauda, C. gracilis, C. limpidus limpidus, C. noxius and C. sculpturatus.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service