Skip to Content
Merck
All Photos(2)

Key Documents

E9138

Corning® reference electrode

Calomel reference electrode

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800

size

1/2 in. (1.3 cm) × 5.1 in. (12.8 cm)

Looking for similar products? Visit Product Comparison Guide

General description

Both feature • Plastic body • Pin-tip connector • 39.5in (99cm) lead
Electrode with replaceable junction, recommended for use in redox titrations, solutions containing suspended solids, Tris buffers.
Corning No. 476406
Fill Solutions: Saturated KCl

Application

For use with pH electrodes and ion-selective electrodes

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service