Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.26
UNSPSC Code:
12352202
Pricing and availability is not currently available.
form
dry concentrate
Quality Segment
usage
sufficient for 50 tests
sensitivity measuring range
0.05-0.1 EU/mL
storage temp.
−20°C
Other Notes
Reconstitute to 5 mL per vial.
Legal Information
E-Toxate is a trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under E-Toxate™ reagent from Limulus polyphemus
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.