Skip to Content
Merck

Skip To

E8512

Ethylmorphine

Synonym(s):

Morphine 3-ethyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H23NO3
CAS Number:
Molecular Weight:
313.39
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
200-970-7
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


drug control

USDEA Schedule II; Home Office Schedule 2; stupéfiant (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal); Kábítószer / Narcotic Drug (Hungary), 78/2022. (XII. 28.) BM rendelet, kontrollierte Droge in Deutschland

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

storage temp.

2-8°C

SMILES string

CCOc1ccc2C[C@@H]3C4C=C[C@H](O)[C@@H]5Oc1c2[C@]45CCN3C

InChI

1S/C19H23NO3/c1-3-22-15-7-4-11-10-13-12-5-6-14(21)18-19(12,8-9-20(13)2)16(11)17(15)23-18/h4-7,12-14,18,21H,3,8-10H2,1-2H3/t12-,13+,14-,18-,19-/m0/s1

InChI key

OGDVEMNWJVYAJL-LEPYJNQMSA-N

Biochem/physiol Actions

Narcotic analgesic; antitussive

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Virginia Tzankova et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 92, 569-579 (2017-06-04)
The toxic liver impairment caused by free radical injury or excessive reactive oxigen species (ROS) formation could be effectivelly attenuated by natural antioxidants. The present study aimed to explore and compare the hepatoprotective and antioxidant effects of free and encapsulated
Arne Helland et al.
Journal of forensic sciences, 55(2), 551-553 (2010-02-18)
Ethylmorphine, an opiate that is partially metabolized to morphine, is a common ingredient in antitussive preparations. We present a case where a 10-month-old boy was administered ethylmorphine in the evening and found dead in bed the following morning. Postmortem toxicological
U D Kuhn et al.
Experimental and toxicologic pathology : official journal of the Gesellschaft fur Toxikologische Pathologie, 50(4-6), 484-490 (1998-10-23)
To investigate whether or not there is a beneficial effect of diltiazem (D) on cyclosporine A (CsA) nephrotoxicity, renal function, CsA blood levels, and effects of CsA on biotransformation in the liver and on lipid peroxidation were characterized in rats.



Questions

Reviews

No rating value

Active Filters