Skip to Content
Merck
All Photos(2)

Key Documents

E7879

Sigma-Aldrich

β-Estradiol 17-acetate

≥99%

Synonym(s):

1,3,5(10)-Estratriene-3,17β-diol 17-acetate, 3,17β-Dihydroxy-1,3,5(10)-estratriene 17-acetate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$198.00
5 G
$770.00

$198.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$198.00
5 G
$770.00

About This Item

Empirical Formula (Hill Notation):
C20H26O3
CAS Number:
Molecular Weight:
314.42
Beilstein:
2625732
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$198.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

sterility

non-sterile

Assay

≥99%

form

powder

solubility

chloroform: 50 mg/mL, clear, colorless to faintly brownish-yellow

shipped in

ambient

storage temp.

room temp

SMILES string

CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C

InChI

1S/C20H26O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h4,6,11,16-19,22H,3,5,7-10H2,1-2H3/t16-,17-,18+,19+,20+/m1/s1

InChI key

QAHOQNJVHDHYRN-SLHNCBLASA-N

Looking for similar products? Visit Product Comparison Guide

Application

β-Estradiol 17-acetate has been used in body burden analyses, to study the effect of triclosan on an estuarine fish.[1]

Biochem/physiol Actions

β-Estradiol is a steroid hormone involved in a number of organ functions. It is significantly associated with brain functions including neuroprotection, neurogenesis and synaptic plasticity.[2] β-Estradiol maintains bone homeostasis, and hence is preferred for osteoporosis therapy.[3] 17-β-Estradiol is the most active estrogen generated in our body.[4] During postmenopausal, 17-β-estradiol inhibits oxidative modification in low density lipoprotein. This indicates that estrogen replacement therapy might be useful in treating cardiovascular disease.[5]

Features and Benefits

This compound is a featured product for ADME Tox research. Click here to discover more featured ADME Tox products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Bioactive Natural Products: Chemistry and Biology, 375-375 (2015)
Layer-by-Layer Films for Biomedical Applications, 517-517 (2014)
Estrogen and the Vessel Wall, 38-38 (2003)
Tomohiro Hikima et al.
European journal of pharmaceutics and biopharmaceutics : official journal of Arbeitsgemeinschaft fur Pharmazeutische Verfahrenstechnik e.V, 54(2), 155-160 (2002-08-23)
We have investigated the distribution of hydrolytic enzymes which metabolize beta-estradiol 17-acetate (EA) to beta-estradiol (E) in man and animal skins in vitro. The distribution of hydrolytic enzymes in human cadaver, hairless dog, rat and hairless mouse skin, was investigated
B H Berg
Biochemistry international, 24(3), 527-533 (1991-06-01)
Adenylyl cyclase activity in the uterus of ovariectomized NMRI mice showed an increase of 87% four minutes after intraperitoneal administration of 17-beta-estradiol 17-beta-acetate in water solution, while in the liver of the same animals activity had decreased by 57%. At

Articles

Discover Bioactive Small Molecules for ADME/Tox

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service