E7409
EPHA3 (571-end), active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
ETK, HEK, TYRO4
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
51111800
NACRES:
NA.32
recombinant
expressed in baculovirus infected Sf9 cells
Quality Level
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
87-120 nmol/min·mg
mol wt
~72 kDa
UniProt accession no.
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... EPHA3(2042)
General description
EPHA3 is a receptor tyrosine kinase (RTK) belonging to the largest subgroup of RTKs called ephrin group of receptors. It has a high level of expression from placental stage through the whole developmental process. It is a type I transmembrane protein, and has a membrane-bound ligand. It is both an oncogene and a tumor-suppressor gene. This protein resides in both the cytosol and the plasma membrane.
Biochem/physiol Actions
EPHA3 binds the ephrin-A ligand and has diverse cellular function. Analysis of human colorectal, breast, lung and pancreatic cancer samples shows somatic mutations in the EPHA3 gene. EPHA3 gene expression can be regulated by CD28 and IGF-1 in Jurkat cells and expression of EPHA3 is associated with adherence and motility of malignant T cells. It maintains the less differentiated state of cancer cells, by activating MAPK (mitogen-activated protein kinase) pathway. It is over-expressed in glioblastoma (GBM) cells, especially during the initial stages. It also has higher expression levels in aggressive mesenchymal subtype of GBM. In hepatocellular carcinoma (HCC), it predicts increased invasiveness and poor patient prognosis.
Physical form
Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.25 mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Cheng-Yi Lu et al.
Oncology reports, 30(5), 2179-2186 (2013-08-24)
Although EphA3 expression has been associated with progression or prognosis in several types of tumors, the role of EphA3 in hepatocellular carcinoma (HCC) remains unknown. This study sought to investigate the clinicopathological and prognostic relevance of EphA3 expression in HCC
Jenni Lahtela et al.
Cell cycle (Georgetown, Tex.), 12(4), 625-634 (2013-01-18)
Activation of a cellular senescence program is a common response to prolonged oncogene activation or tumor suppressor loss, providing a physiological mechanism for tumor suppression in premalignant cells. The link between senescence and tumor suppression supports the hypothesis that a
Structural recognition of an optimized substrate for the ephrin family of receptor tyrosine kinases.
Tara L Davis et al.
The FEBS journal, 276(16), 4395-4404 (2009-08-15)
Ephrin receptor tyrosine kinase A3 (EphA3, EC 2.7.10.1) is a member of a unique branch of the kinome in which downstream signaling occurs in both ligand- and receptor-expressing cells. Consequently, the ephrins and ephrin receptor tyrosine kinases often mediate processes
Laura D Wood et al.
Human mutation, 27(10), 1060-1061 (2006-08-31)
Tyrosine kinases are major regulators of signal transduction cascades involved in cellular proliferation and have important roles in tumorigenesis. We have recently analyzed the tyrosine kinase gene family for alterations in human colorectal cancers and identified somatic mutations in seven
Loraine M Smith et al.
Experimental cell research, 292(2), 295-303 (2003-12-31)
Stimulation of CD28 alone has been shown to regulate cytokine gene transcription and expression of the type 1 insulin-like growth factor receptor (IGF-1R) in lymphocytes. In this study, the ephrin receptor tyrosine kinase ephA3, was identified as a new CD28-responsive
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service