Skip to Content
Merck
All Photos(1)

Key Documents

E7265

Sigma-Aldrich

Big Endothelin 39 rat

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
$695.00

$695.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
0.1 MG
$695.00

About This Item

Empirical Formula (Hill Notation):
C192H292N50O58S5
CAS Number:
Molecular Weight:
4389.00
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.32

$695.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

Gene Information

rat ... Edn1(24323)

Amino Acid Sequence

Cys-Ser-Cys-Ser-Ser-Leu-Met-Asp-Lys-Glu-Cys-Val-Tyr-Phe-Cys-His-Leu-Asp-Ile-Ile-Trp-Val-Asn-Thr-Pro-Glu-Arg-Val-Val-Pro-Tyr-Gly-Leu-Gly-Ser-Pro-Ser-Arg-Ser [Disulfide Bridges: 1-15; 3-11]

General description

Big endothelin-1 is a 39 amino acid peptide, which is synthesized and secreted by vascular smooth muscle and endothelium cells. It is produced as an inactive enzyme, and is cleaved and activated by serine and aspartic proteases and metalloproteinases. Activated form is called ET-1, which is a potent vasoconstrictor. ET-1 is also produced by the action of endothelin converting enzyme (ECE) on big-ET-1. In guinea pigs, big-ET-1 causes the release of prostanoids and thromboxanes.[1][2][3]

Application

Big Endothelin 39 rat has been used for charting of a standard curve in the study of the role of ET-1 in ventricular dilatation.[4]

Biochem/physiol Actions

Precursor of Endothelin-1; vasoconstrictor activity appears to be due to its conversion to endothelin 1 by endothelin converting enzyme in the tissue.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 1 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Fukushige et al.
The Anatomical record, 259(4), 437-445 (2000-07-21)
Immunocytochemical localization of big endothelin-1 (big ET-1), ET-1, and ET receptor A and B (ET(A) and ET(B)), and gene expression of prepro ET-1 mRNA were examined on the rat liver vasculature. Immunoreactivities for big ET-1 and ET-1 were preferentially seen
S Lehoux et al.
British journal of pharmacology, 107(4), 996-1000 (1992-12-01)
1. Changes in vascular permeability following intravenous injections of human big-endothelin-1 (big-ET-1) and endothelin-1 (ET-1) were measured by extravasation of Evans blue dye (EB, 20 mg kg-1) in selected tissues. 2. A low dose of big-ET-1 (40 pmol kg-1) failed
C Fernandez-Patron et al.
Circulation research, 85(10), 906-911 (1999-11-13)
Matrix metalloproteinase-2 (MMP-2, gelatinase A) and its tissue inhibitor (TIMP-2) are mainly known for their roles in the (patho)physiological remodeling of the vasculature, angiogenesis, tissue repair, tumor invasion, inflammation, and atherosclerotic plaque rupture. A mechanism of action of MMP-2 is
D Fraccarollo et al.
Circulation, 96(11), 3963-3973 (1997-12-24)
Left ventricular dilatation after myocardial infarction is associated with impaired ventricular function and heart failure and has important implications for survival. The purpose of this study was to investigate the role of endothelin-1 (ET-1) in ventricular dilatation and the effects
T Sakurai et al.
Biochemical and biophysical research communications, 175(1), 44-47 (1991-02-28)
We report the cloning of a full-length cDNA encoding rat preproendothelin-1 (preproET-1). The predicted rat preproET-1 consists of 202 amino acid residues and highly similar to human, porcine and bovine preproET-1, respectively. The deduced 21-residue sequence of mature rat ET-1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service