Skip to Content
Merck
All Photos(1)

Key Documents

E7159

Sigma-Aldrich

Exo2

≥98% (HPLC)

Synonym(s):

2-Methoxy-4-[(5,6,7,8-tetrahydro-benzo[4,5]thieno[2,3-d]pyrimidin-4-yl)-hydrazonomethyl]-phenol

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$115.00
25 MG
$358.00

$115.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$115.00
25 MG
$358.00

About This Item

Empirical Formula (Hill Notation):
C18H18N4O2S
CAS Number:
Molecular Weight:
354.43
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$115.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

COc1cc(ccc1O)\C=N\Nc2ncnc3sc4CCCCc4c23

InChI

1S/C18H18N4O2S/c1-24-14-8-11(6-7-13(14)23)9-21-22-17-16-12-4-2-3-5-15(12)25-18(16)20-10-19-17/h6-10,23H,2-5H2,1H3,(H,19,20,22)/b21-9+

InChI key

VFNUTEMVQGLDAG-ZVBGSRNCSA-N

Biochem/physiol Actions

Exo2 acts as a chemical probe of intracellular transport.
Exo2 acts as a chemical probe of intracellular transport. Edxo2 has activities similar to Brefeldin A with membrane trafficking to the Golgi appartus, but appears more selective.
Exo2 is a bioactive small molecule that activates Golgi-endoplasmic reticulum (ER) fusion in mammals by distorting the Golgi apparatus. It prevents the release of secretory cargo from the ER. Exo2 do not modify the morphology of the trans-Golgi network (TGN).[1] It also inhibits the anterograde movement of vesicular stomatitis virus G (VSV G) glycoprotein, from ER to Golgi.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lexiang Ji et al.
The Plant cell, 31(10), 2315-2331 (2019-08-24)
Somatic embryogenesis is an important tissue culture technique that sometimes leads to phenotypic variation via genetic and/or epigenetic changes. To understand the genomic and epigenomic impacts of somatic embryogenesis, we characterized soybean (Glycine max) epigenomes sampled from embryos at 10
Ayaka Hieno et al.
Plant physiology, 180(3), 1629-1646 (2019-05-09)
Hydrogen peroxide (H2O2) is a common signal molecule initiating transcriptional responses to all the known biotic and abiotic stresses of land plants. However, the degree of involvement of H2O2 in these stress responses has not yet been well studied. Here
Liwei Lang et al.
Journal of experimental & clinical cancer research : CR, 36(1), 112-112 (2017-08-24)
Although major improvements have been made in surgical management, chemotherapeutic, and radiotherapeutic of prostate cancer, many prostate cancers remain refractory to treatment with standard agents. Therefore, the identification of new molecular targets in cancer progression and development of novel therapeutic
Robert A Spooner et al.
The Biochemical journal, 414(3), 471-484 (2008-06-05)
The small-molecule inhibitor Exo2 {4-hydroxy-3-methoxy-(5,6,7,8-tetrahydrol[1]benzothieno[2,3-d]pyrimidin-4-yl)hydraz-one benzaldehyde} has been reported to disrupt the Golgi apparatus completely and to stimulate Golgi-ER (endoplasmic reticulum) fusion in mammalian cells, akin to the well-characterized fungal toxin BFA (brefeldin A). It has also been reported that
Gertrud Wiedemann et al.
The Plant cell, 30(3), 717-736 (2018-03-09)
RecQ DNA helicases are genome surveillance proteins found in all kingdoms of life. They are characterized best in humans, as mutations in RecQ genes lead to developmental abnormalities and diseases. To better understand RecQ functions in plants we concentrated on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service