Skip to Content
Merck
All Photos(1)

Key Documents

E7029

Sigma-Aldrich

pBICEP-CMV-3 Expression Vector

Sign Into View Organizational & Contract Pricing

Select a Size

20 μG
$1,060.00

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
20 μG
$1,060.00

About This Item

UNSPSC Code:
12171500
NACRES:
NA.56

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

shipped in

dry ice

storage temp.

−20°C

General description

The pBICEP-CMV-3 bicistronic expression vector is a 5.3 kb derivative of pCMV5[1] used for transient or stable co-expression of an N-terminal 1´ FLAGÒ fusion protein and a selection gene in mammalian cells.

Application

For transient, cytoplasmic expression of an N-terminal FLAG® fusion and a second gene of interest or selection marker from bicistronic mRNA. Two genes can be cloned into MCS1 and MCS2 for transcription of a single message from the CMV promoter. This vector is useful for protein-protein interaction studies, multi-subunit proteins, and cloning a selection marker of choice.

Biochem/physiol Actions

The presence of two multiple cloning sites not only allows the user to clone a target gene, but a selectable marker of their choice, as well. The promoter-regulatory region of the human cytomegalovirus immediate early promoter [2][3] drives transcription of the FLAG-fusion construct along with a downstream selection gene. The EMCV IRES [4][5] region controls translation of the selection gene by recruiting the ribosomal subunits for cap-independent translational initiation. Depending on the mode of selection, stable transfectants can be generated by transfection using the appropriate ESCORTä product line specific for the cell type.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pBICEP-CMV is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Jackson et al.
Trends in biochemical sciences, 15(12), 477-483 (1990-12-01)
The initiation of translation of picornaviral RNAs takes place by an unusual mechanism whereby ribosomes bind directly to an internal site rather than scan the RNA from the 5'-end. This internal entry mechanism requires a 450-nucleotide segment of the picornavirus
S K Jang et al.
Journal of virology, 62(8), 2636-2643 (1988-08-01)
Picornavirus RNAs are uncapped messengers and have unusually long 5' nontranslated regions (5'NTRs) which contain many noninitiating AUG triplets. The translational efficiency of different picornavirus RNAs varies between different cell-free extracts and even in the same extract, such as micrococcal
P L Hallauer et al.
BMC genetics, 1, 1-1 (2000-10-19)
Versatile transgenic manipulation of skeletal muscle requires knowledge of the expression profiles of diverse promoter/enhancer elements in the transcriptionally specialized fiber types of which muscle is composed. "Universal" viral promoters/enhancers, e.g., cytomegalovirus IE1 (CMV IE1), are of interest as reagents
S Andersson et al.
The Journal of biological chemistry, 264(14), 8222-8229 (1989-05-15)
The conversion of cholesterol into bile acids in the liver represents the major catabolic pathway for the removal of cholesterol from the body. In this complex biosynthetic pathway, at least 10 enzymes modify both the ring structure and side chain
D R Thomsen et al.
Proceedings of the National Academy of Sciences of the United States of America, 81(3), 659-663 (1984-02-01)
The DNA templates containing immediate early (IE) genes of human cytomegalovirus (CMV) were transcribed in vitro by using a HeLa cell extract. When IE region 1, 2, and 3 were used, transcription was detected qualitatively only from IE region 1.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service