Skip to Content
Merck
All Photos(1)

Documents

E6405

Sigma-Aldrich

Anti-Endothelial Cell Protein C Receptor antibody, Rat monoclonal

clone RCR-379, purified from hybridoma cell culture

Synonym(s):

Anti-EPCR, Monoclonal Anti-Endothelial Cell Protein C Receptor antibody produced in rat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

RCR-379, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: 5-20 μg/mL using HUVEC cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PROCR(10544)

General description

Monoclonal Anti-endothelial cell protein C receptor (EPCR) (rat IgG2a isotype) is derived from the RCR379 hybridoma produced by the fusion of mouse SP2/0 myeloma cells and lymphatic cells isolated from the superficial inguinal lymph nodes from Wister rats immunized with human EPCR-positive RE-1 cells. Endothelial protein C receptor (EPCR), also known as activated protein C receptor (APC receptor), is a protein encoded by the PROCR gene in humans and is mapped to the locus 20q11.22. EPCR is a type 1 transmembrane glycoprotein containing two domains in the extracellular region that are homologous to the α1 and α2 domains of CD1/ major histocompatibility complex class 1 (MHC) class 1 molecules. It is expressed exclusively in endothelial cells and small vessels such as capillaries of the alveolar wall in the lung.

Specificity

Monoclonal Anti-Endothelial Cell Protein C Receptor (EPCR) recognizes human EPCR (49 kDa).

Immunogen

human EPCR-positive RE-1 cells.

Application

Monoclonal Anti-Endothelial Cell Protein C Receptor antibody produced in rat is suitable for flow cytometry at a concentration of 5-20μg/mL using HUVEC cells.
Monoclonal Anti-Endothelial Cell Protein C Receptor antibody produced in rat may be used in immunohistochemistry.

Biochem/physiol Actions

Endothelial protein C receptor (EPCR) also known as activated protein C receptor (APC receptor) is involved in regulation of the cytoprotective and anticoagulant pathways of protein C. EPCR is a member of the CD1/major histocompatibility complex superfamily and plays an important role in regulating the inflammatory response. It is also identified as an endothelial receptor for specific P. falciparum erythrocyte membrane protein 1 (PfEMP1) subtypes. EPCR is associated with an increased risk of venous thromboembolism (VTE).
The EPCR protein also performs other functions, like binding to proteinase. The complex then binds to macrophage 1 antigen (MAC-1) on activated neutrophils and inhibits their adhesion to activated endothelium

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For prolonged storage, freeze in working aliquots at −20 °C. Repeated freezing and thawing is not recom-mended. Storage in frost-free freezers is also not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vaheh Oganesyan et al.
The Journal of biological chemistry, 277(28), 24851-24854 (2002-05-30)
The endothelial cell protein C receptor (EPCR) shares approximately 20% sequence identity with the major histocompatibility complex class 1/CD1 family of molecules, accelerates the thrombin-thrombomodulin-dependent generation of activated protein C, a natural anticoagulant, binds to activated neutrophils, and can undergo
Izumi Naka et al.
Malaria journal, 13, 105-105 (2014-03-19)
Cytoadhesion of Plasmodium falciparum-infected erythrocytes to endothelial cells in microvessels is a remarkable characteristic of severe malaria. The endothelial protein C receptor (EPCR), encoded by the endothelial protein C receptor gene (PROCR), has recently been identified as an endothelial receptor
Z Laszik et al.
Circulation, 96(10), 3633-3640 (1997-12-13)
The protein C anticoagulant pathway is critical to the control of hemostasis. Thrombomodulin and a newly identified receptor for protein C/activated protein C, EPCR, are both present on endothelium. EPCR augments activation of protein C by the thrombin-thrombomodulin complex. To
P Medina et al.
Arteriosclerosis, thrombosis, and vascular biology, 34(3), 684-690 (2014-01-18)
To confirm the effect of the endothelial protein receptor gene (PROCR) haplotypes H1 and H3 on venous thromboembolism (VTE), to study their effect on endothelial protein C receptor (EPCR) expression in human umbilical vein endothelial cells, and to investigate the
Fan Liu et al.
Human genetics, 134(8), 823-835 (2015-05-13)
In the International Visible Trait Genetics (VisiGen) Consortium, we investigated the genetics of human skin color by combining a series of genome-wide association studies (GWAS) in a total of 17,262 Europeans with functional follow-up of discovered loci. Our GWAS provide

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service