Skip to Content
Merck
All Photos(1)

Key Documents

E5155

Sigma-Aldrich

pT7 MAT-Tag® FLAG®-1 Expression Vector

0.5 mg/mL

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

concentration

0.5 mg/mL

shipped in

dry ice

storage temp.

−20°C

Application

Cytoplasmic expression of N-terminal MAT-Tag®-FLAG dual tagged fusion proteins under the T7/lac promoter.

Other Notes

Supplied with a pT7-FLAG-MAT-1+BAP Control Vector.
Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
MAT-Tag is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG-MAT is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Bacterial Expression Vectors: T7 Promoter System. T7 Vectors for Highest Expression Levels in Bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service