Skip to Content
Merck
All Photos(2)

Key Documents

E5000

Sigma-Aldrich

16α,17α-Epoxypregnenolone acetate

~95%

Synonym(s):

16α,17-Epoxy-3β-hydroxy-5-pregnen-20-one acetate, 3β-Acetoxy-16α,17α-epoxy-5-pregnen-20-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H32O4
CAS Number:
Molecular Weight:
372.50
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Assay

~95%

storage temp.

−20°C

SMILES string

CC(=O)OC1CCC2(C)C3CCC4(C)C(C[C@H]5O[C@@]45C(C)=O)C3CC=C2C1

InChI

1S/C23H32O4/c1-13(24)23-20(27-23)12-19-17-6-5-15-11-16(26-14(2)25)7-9-21(15,3)18(17)8-10-22(19,23)4/h5,16-20H,6-12H2,1-4H3/t16?,17?,18?,19?,20-,21?,22?,23-/m1/s1

InChI key

YRLVTBXOGJMZNC-GJCICFDPSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chara Simitzi et al.
Frontiers in bioengineering and biotechnology, 8, 816-816 (2020-08-11)
Mesenchymal stromal cells (MSC) hold significant potential for tissue engineering applications. Modular tissue engineering involves the use of cellularized "building blocks" that can be assembled via a bottom-up approach into larger tissue-like constructs. This approach emulates more closely the complexity
Nina Parmar et al.
Tissue engineering. Part C, Methods, 21(4), 404-412 (2014-09-11)
Muscle degeneration is a prevalent disease, particularly in aging societies where it has a huge impact on quality of life and incurs colossal health costs. Suitable donor sources of smooth muscle cells are limited and minimally invasive therapeutic approaches are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service