Skip to Content
Merck
All Photos(1)

Key Documents

E4274

Sigma-Aldrich

Endotoxin Removal solution

Remove lipopolysaccharides (LPS) from plasmid DNA preparations

Synonym(s):

ET removal buffer, ET removal reagent, ET removal solution, endo removal buffer, endo removal reagent, endo removal solution, endotoxin contamination removal, endotoxin removal buffer, endotoxin removal reagent, plasmid purification endo, plasmid purification endo free

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$285.00

$285.00


Estimated to ship onMay 22, 2025



Select a Size

Change View
25 ML
$285.00

About This Item

UNSPSC Code:
12352200
NACRES:
NA.52

$285.00


Estimated to ship onMay 22, 2025


Quality Level

Application

Endotoxin Removal solution has been used to remove lipopolysaccharide (LPS) from Baker′s yeast RNA.[1]

Biochem/physiol Actions

The reagent relies on the micellar properties of Triton X-114 to remove endotoxins from solution. When warmed above 20 °C, micelles aggregate into a separate phase that sequesters amphiphilic solutes, including lipopolysaccharides. The hydrophilic DNA remains in the aqueous phase.

Quantity

1 volume treats 5 volumes of DNA solution (≤1mg/ml)

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of endotoxin through an aptamer-based impedance biosensor.
Su W
Biosensors And Bioelectronics, 32(1), 32-36 (2012)
M Cotten et al.
Gene therapy, 1(4), 239-246 (1994-07-01)
Endotoxin (lipopolysaccharide, LPS) is commonly found as a contaminant in plasmid DNA preparations. We demonstrate here that the quantities of LPS typically contaminating DNA preparations can generate a toxicity to primary cells (primary human skin fibroblasts, primary human melanoma cells)

Articles

FAQs on bacterial endotoxin contamination, details on endotoxin testing using the LAL assay, and tips to avoid contamination in cell cultures.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service