Key Documents
E4265
Sigma-Aldrich
Embedding molds
26 Flat bullet-shaped cavities double-indexed with numbers and letters, maroon
Select a Size
1 EA
$77.60
$77.60
Estimated to ship onMay 23, 2025
Select a Size
Change View
1 EA
$77.60
About This Item
UNSPSC Code:
41116155
NACRES:
MA.01
$77.60
Estimated to ship onMay 23, 2025
material
silicone
color
maroon
General description
With cavities for the preparation of microscopy samples. Two variations are available. Maroon molds contain additives for chemical and thermal resistance; they are durable and economical. Clear molds are used where optical clarity is necessary for precise sample alignment; recommended for use with low-temperature UV-cured resins.
Choose from one of the most recent versions:
Certificates of Analysis (COA)