Skip to Content
Merck

Skip To

E3389

Epiamastatin hydrochloride

≥97% (HPLC)

Synonym(s):

[(2R,3R)-3-Amino-2-hydroxy-5-methylhexanoyl]-Val-Val-Asp hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C21H38N4O8
CAS Number:
Molecular Weight:
474.55
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352202
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

assay

≥97% (HPLC)

form

powder

storage temp.

−20°C

SMILES string

Cl[H].CC(C)C[C@@H](N)[C@@H](O)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](C(C)C)C(=O)N[C@@H](CC(O)=O)C(O)=O

InChI

1S/C21H38N4O8.ClH/c1-9(2)7-12(22)17(28)20(31)25-16(11(5)6)19(30)24-15(10(3)4)18(29)23-13(21(32)33)8-14(26)27;/h9-13,15-17,28H,7-8,22H2,1-6H3,(H,23,29)(H,24,30)(H,25,31)(H,26,27)(H,32,33);1H/t12-,13+,15+,16+,17-;/m1./s1

InChI key

GBDUPCKQTDKNLS-TXUMRZAQSA-N

Biochem/physiol Actions

Epimer of amastatin at the 2-position. There are no reports of aminopeptidase inhibition by this epimer; on the contrary, there are suggestions that a 2S-hydroxyl group is important for the stability of an initial collision complex with the enzyme.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



D H Rich et al.
Journal of medicinal chemistry, 27(4), 417-422 (1984-04-01)
Amastatin [(2S,3R)-3-amino-2-hydroxy-5-methylhexanoyl-L-valyl-L-valyl-L- aspartic acid] and bestatin [(2S,3R)-3-amino-2-hydroxy-4-phenylbutanoyl-L-leucine] are slow-binding, competitive inhibitors of aminopeptidase M (AP-M) with net inhibition constants (Ki) of 1.9 X 10(-8) and 4.1 X 10(-6) M, respectively. The effect of inhibitor structure on net Ki and on



Global Trade Item Number

SKUGTIN
E3389-1MG04061822415362
E3389-5MG04061833420560

Questions

Reviews

No rating value

Active Filters