Skip to Content
Merck
All Photos(2)

Key Documents

E3156

Sigma-Aldrich

Anti-AtEB1b (C-terminal) antibody produced in rabbit

~2 mg/mL, affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa (additional band at 50 kDa representing degradation products)

species reactivity

Arabidopsis sp.

concentration

~2 mg/mL

technique(s)

western blot: 1-2 μg/mL using leaves extract (cytosolic fraction) of transgenic Arabidopsis seedlings expressing YFP-tagged AtEB1b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Arabidopsis thaliana ... ATEB1A(823923) , ATEB1B(836370)

Related Categories

Specificity

Recognizes AtEB1b fusion protein expressed in transgenic Arabidopsis plants.

Immunogen

synthetic peptide corresponding to amino acids 205-221 located in a region near the C-terminus of Arabidopsis AtEB1b, conjugated to KLH. This sequence is highly conserved (single amino acid substitution) in AtEB1a and AtEB1c.

Application

Anti-AtEB1b (C-terminal) antibody produced in rabbit is suitable for western blotting at a concentration of 1-2μg/mL using leaves extract (cytosolic fraction) of transgenic Arabidopsis seedlings expressing YFP-tagged AtEB1b.

Biochem/physiol Actions

End-binding 1 (EB1) proteins are plus-end-tracking proteins that are specifically localized to growing microtubule plus ends that regulate microtubule and interact with intracellular targets. The plant-specific EB1 subtype functions on the spindle microtubules by accumulating in the prophase nucleus. The microtubule-end-binding protein expressed in Arabidopsis thaliana cells (is called as AtEB1a) is fused with green fluorescent protein (GFP) resulting in labelling of spindle poles. During interphase, AtEB1a-GFP labels the microtubule plus end and also marks the minus end. The microtubule associated protein END BINDING 1b (EB1b) is involved as mutants and dampens root responses to touch and enhances gravitropism. In animals, EB1 proteins have acidic C-terminal tails that might induce an auto-inhibitory conformation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jordi Chan et al.
Nature cell biology, 5(11), 967-971 (2003-10-15)
In plants, it is unclear how dispersed cortical microtubules are nucleated, polarized and organized in the absence of centrosomes. In Arabidopsis thaliana cells, expression of a fusion between the microtubule-end-binding protein AtEB1a and green fluorescent protein (GFP) results in labelling
Shinichiro Komaki et al.
Journal of cell science, 123(Pt 3), 451-459 (2010-01-14)
End-binding 1 (EB1) proteins are evolutionarily conserved plus-end-tracking proteins that localize to growing microtubule plus ends where they regulate microtubule dynamics and interactions with intracellular targets. Animal EB1 proteins have acidic C-terminal tails that might induce an autoinhibitory conformation. Although
Laura Gleeson et al.
Plant science : an international journal of experimental plant biology, 187, 1-9 (2012-03-13)
The ability of roots to navigate around rocks and other debris as they grow through the soil requires a mechanism for detecting and responding to input from both touch and gravity sensing systems. The microtubule associated protein END BINDING 1b

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service