Skip to Content
Merck

E2002

Estriol 3-(β-D-glucuronide) sodium salt

Synonym(s):

1,3,5(10)-Estratriene-3,16α,17β-triol 3-glucuronide, 3,16α,17β-Trihydroxy-1,3,5(10)-estratriene 3-glucuronide

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Linear Formula:
C24H31O9Na
CAS Number:
Molecular Weight:
486.49
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

InChI

1S/C24H32O9.Na.H/c1-24-7-6-13-12-5-3-11(32-23-19(28)17(26)18(27)20(33-23)22(30)31)8-10(12)2-4-14(13)15(24)9-16(25)21(24)29;;/h3,5,8,13-21,23,25-29H,2,4,6-7,9H2,1H3,(H,30,31);;

SMILES string

[Na].CC12CCC3C(CCc4cc(OC5OC(C(O)C(O)C5O)C(O)=O)ccc34)C1CC(O)C2O

InChI key

XGRCPNYENFSILT-UHFFFAOYSA-N

assay

≥98% anhydrous basis (HPLC)

form

powder

solubility

warm water: 20 mg/mL, clear, colorless

shipped in

ambient

storage temp.

−20°C

pictograms

Health hazard

signalword

Warning

hcodes

pcodes

Hazard Classifications

Carc. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Gosland et al.
Cancer research, 53(22), 5382-5385 (1993-11-15)
The multidrug resistance (MDR) gene family has been shown to be highly expressed in several normal tissues including the canalicular membrane of the hepatocyte. We report that a cholestatic estrogen metabolite, 17 beta-estradiol glucuronide (E217G), is a substrate for the
R V Haning et al.
Fertility and sterility, 37(5), 627-632 (1982-05-01)
Twenty-five cycles of induced ovulation with menotropins were investigated blindly with ultrasound to evaluate estrogen monitoring. Plasma 17 beta-estradiol (E2) and urinary estriol glucuronide (E3G) correlated with total ovarian volume (0.58, 0.58), total follicular volume (0.56, 0.52), volume of the
T Nambara et al.
Journal of pharmacobio-dynamics, 6(9), 692-697 (1983-09-01)
The preparation and antigenic properties of estriol 3-glucuronide-bovine serum albumin (BSA) conjugate in which the hapten is linked to the carrier protein through an (O-3-carboxypropylcarbamoylmethyl)oxime bridge at the C-6 position on the steroid nucleus, has been described. 16,17-Di-O-acetyl-6-oxoestriol 6-(O-carboxymethyl)oxime 3-glucuronide
Direct radioimmunoassay of urinary estrogen and pregnanediol glucuronides during the menstrual cycle.
F Z Stanczyk et al.
American journal of obstetrics and gynecology, 137(4), 443-450 (1980-06-15)
I Schiff et al.
American journal of obstetrics and gynecology, 138(8), 1137-1141 (1980-12-15)
A study was designed to compared the metabolic fate and the biologic effects of 4 mg of estriol (E3) administered either orally or vaginally to six postmenopausal women. Blood samples were collected every hour for 6 hours and five different

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service