Skip to Content
Merck
All Photos(1)

Key Documents

E1906

Sigma-Aldrich

Anti-EAPII (TTRAP) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ETS1-associated Protein 2, Anti-TRAF and TNF Receptor Associated Protein

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~41 kDa by calculation

species reactivity

human

technique(s)

western blot: 1:1,000-1:5,000

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... TTRAP(51567)

Immunogen

peptide corresponding to the human EAPII (amino acids 11-28).

Application

Anti-EAPII (TTRAP) antibody produced in rabbit is suitable for western blot analysis at a working dilution of 1:1000-1:5000.

Biochem/physiol Actions

Ets proteins are a family of conserved sequence specific DNA-binding proteins. They function as transcription factors with roles in differentiation, lymphoid cell development, invasiveness, and angiogenesis. ETS1-associated protein II (EAPII) interacts with ETS1 and other Ets proteins (ETS2 and FLI1). EAPII is mainly localized to the nucleus of mammalian cells. EAPII negatively modulates the transcriptional activity of ETS1. It reduces synergistic transactivation by ETS1 and AP-1. Re-expression of EAPII prevents the migration of epithelial cancer cells. It does not affect cell viability.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huiping Pei et al.
Oncogene, 22(18), 2699-2709 (2003-05-14)
Ets proteins constitute a family of conserved sequence-specific DNA-binding proteins and function as transcription factors. ETS1 plays important roles in differentiation, lymphoid cell development, invasiveness and angiogenesis. Such diverse roles of ETS1 are likely to be dependent on its associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service