Skip to Content
Merck
All Photos(1)

Key Documents

E1530

Sigma-Aldrich

Anti-phospho-Epidermal Growth Factor Receptor (pTyr1086) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-phospho-EGFR (pTyr1086)

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous glycerol solution

mol wt

antigen ~185 kDa

species reactivity

human, mouse, rat

technique(s)

western blot: 1:1,000

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr1086)

Gene Information

human ... EGFR(1956)
mouse ... Egfr(13649)

Immunogen

synthetic phosphopeptide derived from the region of EGFR that contains tyrosine 1086.

Biochem/physiol Actions

EGFR is a transmembrane glycoprotein that binds to the EGF family. It is a pivotal regulator of cell proliferation, growth, differentiation and DNA repair after radiation-induced damage. Mutations in the gene encoding EGFR are associated with a number of malignancies including lung cancer and colorectal cancer. Tyr1086 is a minor autophosphorylation site and enables the internalization of the receptor.

Physical form

Solution in Dulbecco′s phosphate buffered saline without Mg2+ and Ca2+, pH 7.3 with 50% glycerol, 1 mg/mL BSA (protease and IgG free) and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael Wolff et al.
Experimental cell research, 317(1), 42-50 (2010-09-23)
The question whether epidermal growth factor (EGF)-induced receptor endocytosis requires the prior autophosphorylation via the EGF receptor (EGFR) kinase domain has been a matter of long-standing debate. In the airway epithelial cell line NCI-H292, the EGFR kinase domain inhibitor BIBW
Jing Bai et al.
Asian Pacific journal of cancer prevention : APJCP, 13(10), 4879-4881 (2012-12-19)
Epidermal growth factor receptor (EGFR) overexpression is associated with resistance to chemotherapy and radiotherapy. The EGFR modulates DNA repair after radiation-induced damage through an association with the catalytic subunit of DNA protein kinase. DNA double-strand breaks (DSBs) are the most
Thu H Truong et al.
Biochemistry, 51(50), 9954-9965 (2012-11-29)
Epidermal growth factor receptor (EGFR) exemplifies the family of receptor tyrosine kinases that mediate numerous cellular processes, including growth, proliferation, and differentiation. Moreover, gene amplification and EGFR mutations have been identified in a number of human malignancies, making this receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service