Skip to Content
Merck
All Photos(1)

Documents

E1282

Sigma-Aldrich

Anti-Epidermal Growth Factor Receptor antibody produced in goat

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-Egfr, Anti-Erbb, Anti-Errp, Anti-Wa5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
51111800
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

mouse

technique(s)

immunohistochemistry: 5-15 μg/mL
western blot: 0.1-0.25 μg/mL

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

mouse ... Egfr(13649)

Immunogen

purified, NSO cell-derived recombinant mouse epidermal growth factor receptor extracellular domain.

Biochem/physiol Actions

EGFR is a transmembrane glycoprotein that binds to the EGF family. It is a pivotal regulator of cell proliferation, growth, differentiation and DNA repair after radiation-induced damage. Mutations in the gene encoding EGFR are associated with a number of malignancies including lung cancer and colorectal cancer.

Physical form

Lyophilized from a 0.2 μm filtered solution in phosphate buffered saline with 5% trehalose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tai Hoon Kim et al.
FEBS letters, 585(1), 121-127 (2010-11-26)
Cuticle formation and molting are critical for the development of Caenorhabditis elegans. To understand cuticle formation more clearly, we screened for suppressors in transgenic worms that expressed dominant ROL-6 collagen proteins. The suro-1 mutant, which is mild dumpy, exhibited a
Takahiro Nakamura et al.
The Journal of clinical investigation, 124(1), 385-397 (2013-12-10)
Corneal integrity and transparency are indispensable for good vision. Cornea homeostasis is entirely dependent upon corneal stem cells, which are required for complex wound-healing processes that restore corneal integrity following epithelial damage. Here, we found that leucine-rich repeats and immunoglobulin-like
Zhilian Li et al.
Diabetes, 67(9), 1847-1857 (2018-07-01)
Previous studies by us and others have indicated that renal epidermal growth factor receptors (EGFR) are activated in models of diabetic nephropathy (DN) and that inhibition of EGFR activity protects against progressive DN in type 1 diabetes. In this study
Ming-Zhi Zhang et al.
Journal of the American Society of Nephrology : JASN, 30(9), 1659-1673 (2019-07-12)
Sex differences mediating predisposition to kidney injury are well known, with evidence indicating lower CKD incidence rates and slower decline in renal function in nondiabetic CKD for premenopausal women compared with men. However, signaling pathways involved have not been elucidated
Yan Li et al.
Diabetes, 70(2), 562-576 (2020-11-27)
Renal epidermal growth factor receptor (EGFR) signaling is activated in models of diabetic nephropathy (DN), and inhibition of the EGFR signaling pathway protects against the development of DN. We have now determined that in cultured podocytes, high glucose led to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service