Skip to Content
Merck

E1007

Ergocalciferol

Sigma Reference Standard

Synonym(s):

Calciferol, Ercalciol, Ergosterol irradiated, Irradiated ergosterol, Vitamin D2

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C28H44O
CAS Number:
Molecular Weight:
396.65
EC Number:
200-014-9
UNSPSC Code:
12352200
PubChem Substance ID:
Beilstein/REAXYS Number:
1916682
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

97.0-103.0% (HPLC)

packaging

ampule of 250 mg

mp

114-118 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)[C@@H](C)/C=C/[C@@H](C)[C@@]1([H])CC[C@@]([C@]1(C)CCC/2)([H])C2=C\C=C(C[C@@H](O)CC3)/C3=C

InChI

1S/C28H44O/c1-19(2)20(3)9-10-22(5)26-15-16-27-23(8-7-17-28(26,27)6)12-13-24-18-25(29)14-11-21(24)4/h9-10,12-13,19-20,22,25-27,29H,4,7-8,11,14-18H2,1-3,5-6H3/b10-9+,23-12+,24-13-/t20-,22+,25-,26+,27-,28+/m0/s1

InChI key

MECHNRXZTMCUDQ-RKHKHRCZSA-N

Gene Information

human ... VDR(7421)

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in flame-sealed ampules under Argon

pictograms

Skull and crossbonesHealth hazard

signalword

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - STOT RE 1 Oral

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sejal Shah et al.
The Journal of adolescent health : official publication of the Society for Adolescent Medicine, 57(1), 19-23 (2015-04-16)
Vitamin D deficiency and cardiometabolic risk factors are common in obese adolescents. Observational studies demonstrate an inverse relationship among serum 25-hydroxyvitamin D (25OHD) and obesity, insulin resistance, and inflammatory cytokines. This pilot study explores if vitamin D supplementation could reduce
R Jorde et al.
European journal of endocrinology, 152(1), 39-45 (2005-03-15)
Smoking is associated with reduced bone density and calcium absorption, and reduced serum levels of vitamin D. A compensatory increase in serum parathyroid hormone (PTH) would therefore be expected as a result of an altered calcium balance. However, reports on
Amanda J Samanek et al.
The Medical journal of Australia, 184(7), 338-341 (2006-04-06)
To examine the influence of geographical and seasonal factors on duration of solar ultraviolet (UV) radiation exposure of skin to produce recommended vitamin D levels without producing erythema. An ecological study using daily Ultraviolet Index (UVI) data collected in major
B L Salle et al.
The American journal of clinical nutrition, 71(5 Suppl), 1317S-1324S (2000-05-09)
During pregnancy, maternal serum concentrations of 25-hydroxyvitamin D, the circulating form of vitamin D, correlate with dietary vitamin D intake. Maternal serum concentrations of 1,25-dihydroxyvitamin D, the hormonal circulating and active form of vitamin D, are elevated during pregnancy; 1,25-dihydroxyvitamin
Pentti Tuohimaa et al.
International journal of cancer, 108(1), 104-108 (2003-11-18)
Vitamin D inhibits the development and growth of prostate cancer cells. Epidemiologic results on serum vitamin D levels and prostate cancer risk have, however, been inconsistent. We conducted a longitudinal nested case-control study on Nordic men (Norway, Finland and Sweden)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service