Skip to Content
Merck
All Photos(2)

Key Documents

E0881

Sigma-Aldrich

Elastatinal

powder

Synonym(s):

(2S)-2-[(4S)-2-Amino-1,4,5,6-tetrahydro4-pyrimidinyl]-N-[[[(1S)-1-carboxy-3-methylbutyl]amino]carbonyl]glycyl-N1-[(1S)-1-methyl-2-oxoethyl-L-glutamamide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$242.00
10 MG
$421.00
25 MG
$858.00

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$242.00
10 MG
$421.00
25 MG
$858.00

About This Item

Empirical Formula (Hill Notation):
C21H36N8O7
CAS Number:
Molecular Weight:
512.56
Beilstein:
873543
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$242.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Elastatinal, microbial

biological source

microbial

form

powder

solubility

water: 20 mg/mL, clear, faintly yellow to yellow

storage temp.

−20°C

SMILES string

[H]C(=O)[C@H](C)NC(=O)[C@H](CCC(N)=O)NC(=O)[C@@H](NC(=O)N[C@@H](CC(C)C)C(O)=O)[C@]1([H])CCNC(=N)N1

InChI

1S/C21H36N8O7/c1-10(2)8-14(19(34)35)28-21(36)29-16(12-6-7-24-20(23)27-12)18(33)26-13(4-5-15(22)31)17(32)25-11(3)9-30/h9-14,16H,4-8H2,1-3H3,(H2,22,31)(H,25,32)(H,26,33)(H,34,35)(H3,23,24,27)(H2,28,29,36)/t11-,12-,13-,14-,16-/m0/s1

InChI key

IJWCGVPEDDQUDE-YGJAXBLXSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Potent, irreversible elastase inhibitor.
Potent, irreversible elastase inhibitor. Elastatinal is a much stronger inhibitor of elastase from pancreas than from leucocytes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

D J Rigby et al.
Biochimica et biophysica acta, 661(2), 315-322 (1981-10-13)
The pyrimidine-3 gene of Neurospora crassa codes for a bifunctional enzyme catalysing the first two steps of the pyrimidine biosynthetic pathway. Difficulties have been experienced in purification due to the lability of the enzyme. The enzyme loses carbamoyl-phosphate synthetase (carbon-dioxide:
The effect of low-molecular-weight inhibitors on the growth in vitro of a human pancreatic adenocarcinoma cell line.
A G Grant et al.
Biochemical Society transactions, 9(1), 134-134 (1981-02-01)
K Marossy et al.
Biochimica et biophysica acta, 615(1), 237-245 (1980-09-09)
The extracts of granules isolated from bovine granulocytes show elastase- and chymotrypsin-like activities, as detected with specific synthetic substrates. Extraction of these enzymes depends upon salt concentration. In the course of the present studies a 21-fold purification of the elastase-like
L Chen et al.
Archives of microbiology, 153(1), 90-94 (1989-01-01)
The effect of the removal of signal peptides after cleavage of precursor molecules by the signal peptidase I was examined in an in vitro translocation system with Escherichia coli membrane vesicles. The translocation of periplasmic alkaline phosphatase precursors was significantly
M Song et al.
Pancreas, 18(3), 231-239 (1999-04-17)
Increased activity of various proteases is observed in both human and experimental pancreatitis; however, the information on the effects of specific protease inhibitors on the disease is limited. In this study we show that a novel elastase inhibitor, guamerin-derived synthetic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service