Key Documents
E0638
Sigma-Aldrich
Horizontal submarine electrophoresis units
gel size 7.5 cm × 10 cm, base unit, dual mode, mini-gel 7.5 cm wide, colorless
About This Item
UNSPSC Code:
41100000
NACRES:
NB.22
material
acrylic
gel size
7.5 cm × 10 cm
color
colorless
General description
Submarine gels may be formed directly on the UV-transparent gel bed using small sliding baffles to contain the agarose as it cools, or outside the unit using a UV-transparent gel tray sealed with tape. Gel is 7.5 cm wide × 10 cm long. Different colors are offered for any color-coding your lab requires (e.g., RNA only).
Combs and tape are not included. Gel tray is not included with the Mini-Gel unit.
Base unit, safety cover, power leads, and all other necessary parts are included.
Choose from one of the most recent versions:
Certificates of Analysis (COA)