Skip to Content
Merck
All Photos(5)

Key Documents

DUO94005

Sigma-Aldrich

Duolink® flowPLA Detection Kit - Violet

Duolink® PLA kit for Flow Cytometry with Violet Detection

Synonym(s):

in situ Proximity Ligation Assay, Flowcytometry-PLA, Protein Protein Interaction Kit

Sign Into View Organizational & Contract Pricing

Select a Size

40 TESTS
$1,060.00

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

Select a Size

Change View
40 TESTS
$1,060.00

About This Item

UNSPSC Code:
41105331
NACRES:
NA.41

$1,060.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onMay 31, 2025


Request a Bulk Order

product line

Duolink®

technique(s)

flow cytometry: suitable
immunofluorescence: suitable
proximity ligation assay: suitable

fluorescence

λex 390 nm; λem 476 nm

suitability

suitable for fluorescence

shipped in

dry ice

storage temp.

−20°C

Application

Specificity

Violet Fluorescence Detection Reagents
Use appropriate laser for ?ex 390 nm excitation
Use appropriate filter for ?em 476 nm emission

Application Note

Primary antibodies are needed. Test your primary antibodies (IgG-class, mono- or polyclonal) in a standard immunofluorescence (IF), immunohistochemistry (IHC), or immunocytochemistry (ICC) assay to determine the optimal fixation, blocking, and titer conditions. Flow validated antibodies are recommended.

Let us do the work for you, learn more about our Custom Service Program to accelerate your Duolink® projects

View full Duolink® product list
Based on proximity ligation assay (PLA), the Duolink® PLA technology allows for endogenous detection of protein interactions, post-translational modifications, and protein expression levels at the single molecule level in fixed cells.

Duolink® flowPLA Detection Kits will enable sensitive detection of proteins, protein-protein interactions, and protein modifications within cell populations by flow cytometry. To perform a Duolink® flowPLA experiment, you will need fixed, suspended cells, two primary antibodies that specifically recognize your proteins of interest, a pair of PLA probes (one 100RXN PLUS and one 100 RXN MINUS), wash buffer and a Duolink® flowPLA Detection Kit. The flowPLA Kits are available with 5 different fluorophores: Violet, Green, Orange, Red, or FarRed. The flowPLA Kits contain all the necessary reagents to perform the amplification and detection of bound PLA probes by flow cytometry. Analysis is carried out using standard flow cytometry assay equipment. User must provide a fixed cell suspension, primary antibodies, and corresponding PLA Probes.

Follow the Duolink® PLA Flow Cytometry Protocol to use this product.

Visit our Duolink® PLA Flow Cytometry page on how to run a Duolink® flow experiment, applications, troubleshooting, and more.

Features and Benefits

  • Analyze protein protein interactions with flow cytometry readout
  • Analyze cell populations with Proximity Ligation Assay
  • Increased sensitivity due to rolling circle amplification for low abundant targets
  • No overexpression or genetic manipulation required
  • Relative quantification possible
  • Works with any flow cytometer instrumentation
  • Easy to follow flexible protocol
  • Publication-ready results

Components

This product is comprised of the following:
  • 5x Detection Solution - Violet (DUO84005)
  • 5x Ligation Buffer (DUO82009)
  • 5x Amplification Buffer (DUO82050)
  • Ligase (1U/μL)
  • Polymerase (10U/μL)

See datasheet for more information.

Legal Information

Duolink is a registered trademark of Merck KGaA, Darmstadt, Germany
PLA is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Considerations for proper experimental design, preparation and execution of the Duolink® PLA for flow cytometry protocol.

General tips and tricks for proper experiment execution, aid in identifying potential problems, and provide solutions to ensure a successful Duolink® PLA experiment for flow cytometry.

Duolink® PLA kit enhances flow cytometry for detecting protein interactions accurately.

Learn how Proximity Ligation Assay technology works and how the protein-protein interaction control kit can confirm in situ detection of EGF-induced EGFR-HER2 dimerization.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service