D9768
Lambda Phage DNA, Methylated from Escherichia coli host strain W3110
lyophilized powder
Select a Size
$72.20
Estimated to ship onMay 29, 2025
Select a Size
About This Item
$72.20
Estimated to ship onMay 29, 2025
Recommended Products
grade
Molecular Biology
for molecular biology
form
lyophilized powder
mol wt
31.5 × 103 kDa
48 kb
suitability
suitable for substrate for restriction endonucleases
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Specificity
Application
Infecting E. coli strain W3110 with lambda C1857 strain creates E. coli lysogen cultures. The phage is released from E. coli cell pellets by lysing with a high salt buffer, pH 8.0. The crude mixture is passed through a series of enzymatic steps, multiple cesium gradients, and phage DNA is dialyzed against 1 mM Tris-HCl, pH 8.0, and 1 mM magnesium chloride. The DNA is finally extracted by phenol-chloroform solution.
Unit Definition
Reconstitution
Incubate the DNA sample vial at 65 °C for 4 hrs with frequent mixing.
After reconstitution, the product should be stored at -20 °C.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service