Key Documents
D9054
Nunc® petri dishes
diam. × H 150 mm × 25 mm, surface area 148 cm2, Deep Petri without stacking ring; non-vented, Lab-Tek® petri dish
Synonym(s):
culture dishes, disposable petri dishes, plastic petri dishes
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
material
polystyrene
round
sterility
sterile
packaging
pack of 12 ea
case of 72 ea (Internal pack 12)
manufacturer/tradename
Nunc 4014-12
diam. × H
150 mm × 25 mm
surface area
148 cm^2
148 cm2
binding type
non-treated surface
General description
Nunc Lab-Tek® and standard petri dishes are optimized for culturing fungi, bacteria and other microorganisms in addition to supporting automated high-throughput (HTS) applications.
  • For culturing of fungi, bacteria and other microorganisms
  • Contact dish useful for sampling in hospital environments (for research use only), as well as in food science and the pharmaceutical industry
  • Perform well in automatic dispensers due to complete flatness and uniform height
  • Available with a moulded grid
  • Deep Petri Dishes allow for longer culture periods
  • Compatible with automated systems
Please note: Due to these dishes being constructed of polystyrene (not glass) the walls of the dish may be thinner; this results in a size difference in the outer diameter of approx 10 mm.
Legal Information
Lab-Tek is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)