Skip to Content
Merck
All Photos(1)

Key Documents

D8816

Sigma-Aldrich

N-(3,3-Diphenylpropyl)glycinamide

>98% (HPLC), solid

Synonym(s):

N20C

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$113.00

$113.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
$113.00

About This Item

Empirical Formula (Hill Notation):
C17H20ON2
CAS Number:
Molecular Weight:
268.35
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.77

$113.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

>98% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 26 mg/mL

storage temp.

−20°C

SMILES string

NC(=O)CNCCC(c1ccccc1)c2ccccc2

InChI

1S/C17H20N2O/c18-17(20)13-19-12-11-16(14-7-3-1-4-8-14)15-9-5-2-6-10-15/h1-10,16,19H,11-13H2,(H2,18,20)

InChI key

BYGXICRJQZKUPA-UHFFFAOYSA-N

Biochem/physiol Actions

NMDA glutamate receptor open channel blocker.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service