Skip to Content
Merck
All Photos(1)

Key Documents

D7662

Supelco

Delorazepam

Synonym(s):

7-Chloro-5-(o-chlorophenyl)-1,3-dihydro-2H-1,4-benzodiazepin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10Cl2N2O
CAS Number:
Molecular Weight:
305.16
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:

drug control

USDEA Schedule IV; Home Office Schedule 4.1; psychotrope (France); kontrollierte Droge in Deutschland; regulated under CDSA - not available from Sigma-Aldrich Canada

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

Clc1ccc2NC(=O)CN=C(c3ccccc3Cl)c2c1

InChI

1S/C15H10Cl2N2O/c16-9-5-6-13-11(7-9)15(18-8-14(20)19-13)10-3-1-2-4-12(10)17/h1-7H,8H2,(H,19,20)

InChI key

CHIFCDOIPRCHCF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Rocca et al.
Acta psychiatrica Scandinavica, 95(5), 444-450 (1997-05-01)
Recently, there has been a renewed interest in alternatives to the benzodiazepines for the treatment of generalized anxiety disorder (GAD). The aim of the present study was to compare the efficacy of paroxetine vs. imipramine and 2'-chlordesmethyldiazepam in 81 patients
J Sennesael et al.
European journal of clinical pharmacology, 41(1), 65-68 (1991-01-01)
The pharmacokinetics of a single 2 mg IV dose of chlordesmethyldiazepam has been studied in 11 patients with renal failure on regular haemodialysis and in 11 age-matched healthy controls. The kinetics was also examined after a single 2 mg oral
S R Bareggi et al.
European journal of clinical pharmacology, 48(3-4), 265-268 (1995-01-01)
We studied the pharmacokinetics of a single 0.5-mg i.v. dose of chlordesmethyldiazepam in 8 patients with liver disease and in 12 age-matched healthy controls. The kinetics were also studied of a single 1-mg oral dose in the patients with liver
Premedication with chlordemethyldiazepam and anxiolytic effect of diazepeam in implantology.
G Manani et al.
Anesthesia progress, 42(3-4), 107-112 (1995-01-01)
[A ketamine-chlordemethyldiazepam combination in celioscopy].
E Struzziero et al.
Minerva ginecologica, 40(12), 715-717 (1988-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service