Skip to Content
Merck
All Photos(3)

Key Documents

D7003

Sigma-Aldrich

DL-Dihydroorotic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H6N2O4
CAS Number:
Molecular Weight:
158.11
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:

form

powder

SMILES string

OC(=O)C1CC(=O)NC(=O)N1

InChI

1S/C5H6N2O4/c8-3-1-2(4(9)10)6-5(11)7-3/h2H,1H2,(H,9,10)(H2,6,7,8,11)

InChI key

UFIVEPVSAGBUSI-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timo Heikkilä et al.
Journal of medicinal chemistry, 50(2), 186-191 (2007-01-19)
Pyrimidine biosynthesis presents an attractive drug target in malaria parasites due to the absence of a pyrimidine salvage pathway. A set of compounds designed to inhibit the Plasmodium falciparum pyrimidine biosynthetic enzyme dihydroorotate dehydrogenase (PfDHODH) was synthesized. PfDHODH-specific inhibitors with
Joana Costeira-Paulo et al.
Cell chemical biology, 25(3), 309-317 (2018-01-24)
The interactions between proteins and biological membranes are important for drug development, but remain notoriously refractory to structural investigation. We combine non-denaturing mass spectrometry (MS) with molecular dynamics (MD) simulations to unravel the connections among co-factor, lipid, and inhibitor binding
Mihwa Lee et al.
Journal of molecular biology, 348(3), 523-533 (2005-04-14)
Escherichia coli dihydroorotase has been crystallized in the presence of the product, L-dihydroorotate (L-DHO), and the structure refined at 1.9A resolution. The structure confirms that previously reported (PDB entry 1J79), crystallized in the presence of the substrate N-carbamyl-D,L-aspartate (D, L-CA-asp)
O Björnberg et al.
Archives of biochemistry and biophysics, 391(2), 286-294 (2001-07-05)
The flavoprotein dihydroorotate dehydrogenase (DHOD) catalyzes the oxidation of dihydroorotate to orotate. Dihydrooxonate is an analogue of dihydroorotate in which the C5 carbon is substituted by a nitrogen atom. We have investigated dihydrooxonate as a substrate of three DHODs, each
Eloi R Verrier et al.
Gut, 69(1), 158-167 (2019-03-06)
Hepatitis D virus (HDV) is a circular RNA virus coinfecting hepatocytes with hepatitis B virus. Chronic hepatitis D results in severe liver disease and an increased risk of liver cancer. Efficient therapeutic approaches against HDV are absent. Here, we combined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service