Skip to Content
Merck
All Photos(2)

Key Documents

D6942

Sigma-Aldrich

DK-AH 269

≥98% (HPLC), solid

Synonym(s):

(S)-(+)-7,8-Dimethoxy-3-[[1-(2-(3,4-dimethoxyphenyl)ethyl)-3-piperidinyl]methyl]-1,3,4,5-tetrahydro-2H-3-benzazepin-2-one hydrochloride, Cilobradine hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
$271.00
25 MG
$1,030.00

$271.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Select a Size

Change View
5 MG
$271.00
25 MG
$1,030.00

About This Item

Empirical Formula (Hill Notation):
C28H38N2O5 · HCl
CAS Number:
Molecular Weight:
519.07
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

$271.00


Estimated to ship onApril 14, 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white

solubility

H2O: ~20 mg/mL

storage temp.

2-8°C

SMILES string

O=C1CC2=CC(OC)=C(OC)C=C2CCN1C[C@@H](CCC3)CN3CCC4=CC(OC)=C(OC)C=C4.[H]Cl

InChI

1S/C28H38N2O5.ClH/c1-32-24-8-7-20(14-25(24)33-2)9-12-29-11-5-6-21(18-29)19-30-13-10-22-15-26(34-3)27(35-4)16-23(22)17-28(30)31;/h7-8,14-16,21H,5-6,9-13,17-19H2,1-4H3;1H/t21-;/m0./s1

InChI key

MTAKUIYCCYKGJJ-BOXHHOBZSA-N

Application

DK-AH 269 has been used as a hyperpolarization-activated and cyclic nucleotide-gated channel (HCN) antagonist to study its effects on the inhibition of Ih in HCN-deficient hair cells of mice.[1] It has also been used as an HCN blocker to study its effects on action potential firing in the suprachiasmatic nucleus (SCN) neurons.[2]

Biochem/physiol Actions

DK-AH 269 (Cilobradine) is an HCN Channel blocker; an open channel blocker of neuronal Ih and related cardiac If channels.
DK-AH 269 is a bradycardiac agent.[3]

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Cyclic Nucleotide-Gated (CNG) and Hyperpolarization Activated Cyclic Nucleotide-Gated (HCN) Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effects of a bradycardic agent (DK-AH 269) on haemodynamics and oxygen consumption of isolated blood-perfused rabbit hearts
Granetzny A, et al.
Journal of Clinical and Basic Cardiology, 3(3), 191-196 (2000)
Vivien Chevaleyre et al.
Proceedings of the National Academy of Sciences of the United States of America, 99(14), 9538-9543 (2002-07-03)
Hyperpolarization-activated nonselective cation channels (Ih channels) play an important role in the control of membrane excitability and rhythmic neuronal activity. The functional relevance of presynaptic Ih channels in regulating synaptic function, however, is not well established. Recently, it has been
H C Pape
Neuroscience, 59(2), 363-373 (1994-03-01)
A class of pharmacologically active substances, known as "specific bradycardic agents", exerts a negative chronotropic influence on cardiac activity, which heavily relies upon a potent blockade of the hyperpolarization-activated cation current in Purkinje fibers. Since the cation conductance activated by
Cyclic AMP signaling control of action potential firing rate and molecular circadian pacemaking in the suprachiasmatic nucleus
Atkinson S E, et al.
Journal of biological rhythms, 26(3), 210-220 (2011)
HCN channels expressed in the inner ear are necessary for normal balance function
Horwitz G C, et al.
The Journal of Neuroscience, 31(46), 16814-16825 (2011)

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service