Skip to Content
Merck

Skip To

D6500

2′-Deoxyadenosine 5′-triphosphate disodium salt

≥97%

Synonym(s):

dATP-Na2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 mg
Check Cart for Availability
$166.00
25 mg
Check Cart for Availability
$265.00
100 mg
Check Cart for Availability
$706.00

About This Item

Empirical Formula (Hill Notation):
C10H14N5Na2O12P3
CAS Number:
Molecular Weight:
535.15
PubChem Substance ID:
eCl@ss:
32160414
UNSPSC Code:
41106305
NACRES:
NA.51
EC Number:
277-809-2
MDL number:
Assay:
≥97%
Biological source:
natural (inorganic)
Form:
powder
Solubility:
water: 50 mg/mL, clear, colorless
Storage temp.:
−20°C

$166.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

natural (inorganic)

Quality Segment

assay

≥97%

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

[Na+].[Na+].Nc1ncnc2n(cnc12)C3C[C@H](O)[C@@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)O3

InChI

1S/C10H16N5O12P3.2Na/c11-9-8-10(13-3-12-9)15(4-14-8)7-1-5(16)6(25-7)2-24-29(20,21)27-30(22,23)26-28(17,18)19;;/h3-7,16H,1-2H2,(H,20,21)(H,22,23)(H2,11,12,13)(H2,17,18,19);;/q;2*+1/p-2/t5-,6+,7?;;/m0../s1

InChI key

JEKDCIBJADJZSK-LNUHRGGJSA-L

Application

2′-Deoxyadenosine 5′-triphosphate (dATP) is used as a substrate by a variety of polymerases including DNA polymerase(s) and reverse transcriptase(s).
2′-Deoxyadenosine 5′-triphosphate disodium salt has been used:
  • for Smac- X-linked inhibitor of apoptosis protein (Smac-XIAP) binding assay.
  • as a component of polymerase chain reaction (PCR) using AM-toxin primers.[1]
  • to determine the affinity constant (Kd) with the transcription termination factor Rho.
  • to determine the activity of poly ADP-ribose polymerase 1 (PAPR-1 ).

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
D4001D6000A26209
biological source

natural (inorganic)

biological source

synthetic (organic)

biological source

synthetic (organic)

biological source

-

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

powder

form

powder

form

powder

form

-

assay

≥97%

assay

≥95%

assay

≥95% (HPLC)

assay

99%

solubility

water: 50 mg/mL, clear, colorless

solubility

-

solubility

-

solubility

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters