Skip to Content
Merck

Skip To

D6035

Disopyramide phosphate salt

Synonym(s):

α-Diisopropylaminoethyl-α-phenylpyridine-2-acetamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
$72.10
10 g
Check Cart for Availability
$233.00

About This Item

Linear Formula:
C21H29N3O · H3PO4
CAS Number:
Molecular Weight:
437.47
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
EC Number:
244-756-1
MDL number:
Form:
solid

$72.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

Quality Segment

SMILES string

OP(O)(O)=O.CC(C)N(CCC(C(N)=O)(c1ccccc1)c2ccccn2)C(C)C

InChI

1S/C21H29N3O.H3O4P/c1-16(2)24(17(3)4)15-13-21(20(22)25,18-10-6-5-7-11-18)19-12-8-9-14-23-19;1-5(2,3)4/h5-12,14,16-17H,13,15H2,1-4H3,(H2,22,25);(H3,1,2,3,4)

InChI key

CGDDQFMPGMYYQP-UHFFFAOYSA-N

Application

Disopyramide phosphate salt has been used as an internal standard for plasma sample analysis by mass spectrometry.[1] It might be used as a test compound in a teratogenicity study.[2]

Biochem/physiol Actions

Disopyramide is found to stabilize the cell membrane by altering the action potential. It might act as an antiarrhythmic agent.
Class IA antiarrhythmic; sodium channel blocker

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
474500178251178196
form

solid

form

solid

form

solid

form

solid

Quality Level

200

Quality Level

100

Quality Level

100

Quality Level

100

Gene Information

human ... SCN10A(6336), SCN11A(11280), SCN1A(6323), SCN2A(6326), SCN3A(6328), SCN4A(6329), SCN5A(6331), SCN7A(6332), SCN8A(6334), SCN9A(6335)

Gene Information

-

Gene Information

-

Gene Information

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters