Skip to Content
Merck
All Photos(1)

Key Documents

D5821

Sigma-Aldrich

DS2

≥98% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H12ClN3OS
CAS Number:
Molecular Weight:
353.83
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥98% (HPLC)

form

powder

color

off-white to light tan

solubility

DMSO: >10 mg/mL

storage temp.

2-8°C

SMILES string

Clc1ccc(cc1)C(=O)Nc2c(nc3ccccn23)-c4cccs4

InChI

1S/C18H12ClN3OS/c19-13-8-6-12(7-9-13)18(23)21-17-16(14-4-3-11-24-14)20-15-5-1-2-10-22(15)17/h1-11H,(H,21,23)

InChI key

AZKMWHRDICVYEI-UHFFFAOYSA-N

Biochem/physiol Actions

DS2 is a selective positive allosteric modulator of δ-GABAA receptors.

Features and Benefits

This compound is featured on the GABAA Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minjun Yang et al.
Blood, 136(8), 946-956 (2020-05-10)
Mutations in the FMS-like tyrosine kinase 3 (FLT3) gene in 13q12.2 are among the most common driver events in acute leukemia, leading to increased cell proliferation and survival through activation of the phosphatidylinositol 3-kinase/AKT-, RAS/MAPK-, and STAT5-signaling pathways. In this
Sjoukje-Marije Haisma et al.
PloS one, 14(4), e0214751-e0214751 (2019-04-19)
Treatment decisions in inflammatory bowel diseases are increasingly based on longitudinal tracking of calprotectin results. Many hospital laboratories measure calprotectin levels in sent-in stool samples with an enzyme-linked immunosorbent assay (ELISA). Several manufacturers introduced a lateral flow-based test with software
Julianne Megaw et al.
Frontiers in microbiology, 8, 1589-1589 (2017-09-06)
Persister cells are phenotypic variants within a microbial population, which are dormant and transiently tolerant to stress. Persistence has been studied extensively in bacteria, and in eukaryotes to a limited extent, however, it has never been observed in archaea. Using
Wing Sun Faith Chung et al.
BMC biology, 14, 3-3 (2016-01-13)
Dietary intake of specific non-digestible carbohydrates (including prebiotics) is increasingly seen as a highly effective approach for manipulating the composition and activities of the human gut microbiota to benefit health. Nevertheless, surprisingly little is known about the global response of
Cynthia L Darnell et al.
mBio, 11(4) (2020-08-14)
Precise control of the cell cycle is central to the physiology of all cells. In prior work we demonstrated that archaeal cells maintain a constant size; however, the regulatory mechanisms underlying the cell cycle remain unexplored in this domain of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service