Skip to Content
Merck

Skip To

D5307

Amplification Grade DNase 1

Synonym(s):

Deoxyribonuclease I (dnase 1) from bovine pancreas

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
1000 units
Check Cart for Availability
$718.00

About This Item

NACRES:
NA.53
UNSPSC Code:
12352204

$718.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

storage temp.

−20°C

Other Notes

amplification grade** unlisted item

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AMPD110104159001260913
form

liquid

form

liquid

form

lyophilized

form

lyophilized

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

-

Quality Level

200


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
4847904061833691229
4817904061838086341
4818204061837619243
4888904061838242334
D5307-1000UN04061832854793
D5307-1KU04061826599402

Questions

Reviews

No rating value

Active Filters