Skip to Content
Merck
All Photos(1)

Key Documents

D5004

Sigma-Aldrich

N,N-Dimethyl-p-phenylenediamine monohydrochloride

Synonym(s):

4-(Dimethylamino)aniline monohydrochloride, 4-Amino-N,N-dimethylaniline monohydrochloride, DMPPDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4NH2·HCl
CAS Number:
Molecular Weight:
172.66
Beilstein:
4546912
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

form

powder

mp

215 °C (dec.) (lit.)

storage temp.

−20°C

SMILES string

Cl[H].CN(C)c1ccc(N)cc1

InChI

1S/C8H12N2.ClH/c1-10(2)8-5-3-7(9)4-6-8;/h3-6H,9H2,1-2H3;1H

InChI key

KTWNIUBGGFBRKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Caution

Darkens readily to grey powder when exposed to air.

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Resp. Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

V Fogliano et al.
Journal of agricultural and food chemistry, 47(3), 1035-1040 (1999-12-20)
A novel method for measuring the antioxidant activity using N, N-dimethyl-p-phenylenediamine (DMPD) was developed. The radical cation of this compound gives a stable colored solution and a linear inhibition of color formation can be observed in the presence of 0.
Veronica Verde et al.
Free radical research, 36(8), 869-873 (2002-11-08)
Oxidative stress has been clearly implicated in human disease by a growing body of scientific evidences. There is no ideal method for the measurement of this parameter. A possible strategy would be to measure simultaneously several biomarkers representing damage to
Kraiwan Punyain et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 368-378 (2011-09-20)
Organic radical cations form dicationic dimers in solution, observed experimentally as diamagnetic species in temperature-dependent EPR and low temperature UV/Vis spectroscopy. Dimerization of paraphenylenediamine, N,N-dimethyl-paraphenylenediamine and 2,3,5,6-tetramethyl-paraphenylenediamine radical cation in ethanol/diethylether mixture was investigated theoretically according to geometry, energetics and
Francisco J Morales et al.
Journal of agricultural and food chemistry, 50(16), 4657-4661 (2002-07-26)
The Maillard reaction has a dramatic impact on the overall acceptance and nutritional quality of most of the foods consumed daily in European countries. Melanoidins are polymeric structures formed in the last stage of the Maillard reaction with nearly unknown
F Tessier et al.
International journal for vitamin and nutrition research. Internationale Zeitschrift fur Vitamin- und Ernahrungsforschung. Journal international de vitaminologie et de nutrition, 66(2), 166-170 (1996-01-01)
An HPLC micro-method with fluorescence detection has been developed to determine total vitamin C (vit C) and dehydroascorbic acid (DHA) concentrations in human plasma samples. This method is based on the rapid, specific reaction of DHA with dimethyl-o-phenylenediamine (DMPD) to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service