Skip to Content
Merck
All Photos(1)

Key Documents

D4813

Sigma-Aldrich

Dendrotoxin-K

≥98% (HPLC), lyophilized powder

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

lyophilized powder

mol wt

6,542 Da

storage temp.

−20°C

Application

Dendrotoxin-K is suitable for use as a potassium voltage-gated channel subfamily A member 1 (Kv1.1) channel blocker in axon and human embryonic kidney (HEK293) cells. It has also been used as a selective blocker of Kv channels in mice.

Biochem/physiol Actions

Dendrotoxin-K (DTXk) is isolated from mamba snake Dendroaspis polylepis and interacts with the β-turn region in the N-terminal sequence of the potassium channel. It inhibits tumor progression in gefitinib-resistant non-small cell lung cancer cells. DTXk induces neuronal damage in the hippocampus via N-methyl-d-aspartate (NMDA) and non-NMDA receptors.
Dendrotoxin-K inhibits potassium channels that contain Kv1.1 protein only.

Features and Benefits

This compound is featured on the Potassium Channels page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Other Notes

Isolated initially from Dendroaspis polylepis polylepis venom

Preparation Note

Purified by a modification of the method of Schweitz.

Reconstitution

Reconstitute in 1 mL deionized water or buffer (pH 7.5) to yield 10 μM stock

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Analog modulation of spike-evoked transmission in CA 3 circuits is determined by axonal K v1. 1 channels in a time-dependent manner
Bialowas A, et al.
The European Journal of Neuroscience, 41(3), 293-304 (2015)
Cav1. 3 calcium channels are required for normal development of the auditory brainstem
Hirtz JJ, et al.
The Journal of Neuroscience, 31(22), 8280-8294 (2011)
F C Wang et al.
European journal of biochemistry, 263(1), 222-229 (1999-08-03)
Dendrotoxin (DTX) homologues are powerful blockers of K+ channels that contain certain subfamily Kv1 (1.1-1.6) alpha- and beta-subunits, in (alpha)4(beta)4 stoichiometry. DTXk inhibits potently Kv1.1-containing channels only, whereas alphaDTX is less discriminating, but exhibits highest affinity for Kv1.2. Herein, the
Effects of voltage-gated K+ channel blockers in gefitinib-resistant H460 non-small cell lung cancer cells
Jeon WIL, et al.
Anticancer Research, 32(12), 5279-5284 (2012)
Identification of residues in dendrotoxin K responsible for its discrimination between neuronal K+ channels containing Kv1. 1 and 1.2 alpha subunits
Wang FC, et al.
European Journal of Biochemistry, 263(1), 222-229 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service