Skip to Content
Merck
All Photos(1)

Key Documents

D4556

Sigma-Aldrich

D-threo-Dihydrosphingosine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H39NO2
CAS Number:
Molecular Weight:
301.51
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCC[C@@H](O)[C@H](N)CO

InChI

1S/C18H39NO2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(21)17(19)16-20/h17-18,20-21H,2-16,19H2,1H3/t17-,18-/m1/s1

InChI key

OTKJDMGTUTTYMP-QZTJIDSGSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiaoya Qin et al.
Plant physiology and biochemistry : PPB, 119, 70-80 (2017-08-29)
The fungal toxin Fumonisin B1 (FB1) is a strong inducer to trigger plant hypersensitive responses (HR) along with increased long chain bases (LCB) and long chain base phosphates (LCBP) contents, though the regulatory mechanism of FB1 action and how the
Yidi Sun et al.
Molecular biology of the cell, 23(12), 2388-2398 (2012-04-27)
Sphingoid intermediates accumulate in response to a variety of stresses, including heat, and trigger cellular responses. However, the mechanism by which stress affects sphingolipid biosynthesis has yet to be identified. Recent studies in yeast suggest that sphingolipid biosynthesis is regulated
Mariana Saucedo-García et al.
The New phytologist, 191(4), 943-957 (2011-05-04)
Long chain bases (LCBs) are sphingolipid intermediates acting as second messengers in programmed cell death (PCD) in plants. Most of the molecular and cellular features of this signaling function remain unknown. We induced PCD conditions in Arabidopsis thaliana seedlings and
Emma M Dangerfield et al.
Chembiochem : a European journal of chemical biology, 13(9), 1349-1356 (2012-05-29)
The immunomodulatory glycolipid α-galactosylceramide (α-GalCer) binds to CD1d and exhibits potent activity as a ligand for invariant CD1d-restricted natural killer-like T cells (iNKT cells). Structural analogues of α-GalCer have been synthesised to determine which components are required for CD1d presentation
Lichao Hu et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 34(5), 2573-2581 (2013-04-26)
Our previous study has demonstrated that tissue factor-factor VIIa (TF/FVIIa) complex promotes the proliferation and migration of colon cancer cell line SW620 through the activation of protease-activated receptor 2 (PAR2). In the current study, the underlying molecular mechanisms of TF/FVIIa/PAR2

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service