Skip to Content
Merck

Skip To

D4130

Barium diphenylamine-4-sulfonate

redox indicator

Synonym(s):

4-(Phenylamino)benzenesulfonic acid barium salt, Diphenylamine-4-sulfonic acid barium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(C12H10NO3S)2Ba
CAS Number:
Molecular Weight:
633.88
NACRES:
NA.21
PubChem Substance ID:
UNSPSC Code:
12161500
EC Number:
228-278-0
MDL number:
Beilstein/REAXYS Number:
3795146
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Barium diphenylamine-4-sulfonate, for redox titration

Quality Level

assay

≥95%

form

powder

technique(s)

titration: suitable

solubility

50 mg/mL, clear to very slightly hazy, faintly yellow-green

cation traces

C: 44.3-46.6%, N: 4.1-4.8%

SMILES string

[Ba++].[O-]S(=O)(=O)c1ccc(Nc2ccccc2)cc1.[O-]S(=O)(=O)c3ccc(Nc4ccccc4)cc3

InChI

1S/2C12H11NO3S.Ba/c2*14-17(15,16)12-8-6-11(7-9-12)13-10-4-2-1-3-5-10;/h2*1-9,13H,(H,14,15,16);/q;;+2/p-2

InChI key

IVCNVXFNTKXMCA-UHFFFAOYSA-L

General description

Visit our Titration Center to learn more.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1.00255242969433373
technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

titration: suitable

technique(s)

-

assay

≥95%

assay

-

assay

-

assay

~95%

cation traces

C: 44.3-46.6%, N: 4.1-4.8%

cation traces

-

cation traces

-

cation traces

-

solubility

50 mg/mL, clear to very slightly hazy, faintly yellow-green

solubility

water: soluble at 20 °C

solubility

-

solubility

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

form

powder

form

solid

form

-

form

powder


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class

11 - Combustible Solids

wgk

WGK 1

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
D4130-50G04061833629567
D4130-25G04061833564059
D4130-5G04061833564066

Questions

Reviews

No rating value

Active Filters