Skip to Content
Merck
All Photos(2)

Key Documents

D3534

Sigma-Aldrich

cis-7,10,13,16-Docosatetraenoic acid methyl ester

≥98% (capillary GC)

Synonym(s):

Methyl 7Z,10Z,13Z,16Z-docosatetraenoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
$296.00

$296.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
25 MG
$296.00

About This Item

Empirical Formula (Hill Notation):
C23H38O2
CAS Number:
Molecular Weight:
346.55
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

$296.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request a Bulk Order

Assay

≥98% (capillary GC)

form

liquid

functional group

ester

lipid type

omega FAs

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCC\C=C/C\C=C/C\C=C/C\C=C/CCCCCC(=O)OC

InChI

1S/C23H38O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23(24)25-2/h7-8,10-11,13-14,16-17H,3-6,9,12,15,18-22H2,1-2H3/b8-7-,11-10-,14-13-,17-16-

InChI key

ABGHYAFHPINIHF-ZKWNWVNESA-N

Related Categories

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service