Skip to Content
Merck
All Photos(1)

Key Documents

D3128

Sigma-Aldrich

DCC

Synonym(s):

N,N′-Dicyclohexylcarbodiimide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11N=C=NC6H11
CAS Number:
Molecular Weight:
206.33
Beilstein:
610662
EC Number:
MDL number:
UNSPSC Code:
12352200

bp

122-124 °C/6 mmHg (lit.)

mp

34-35 °C (lit.)

SMILES string

C1CCC(CC1)N=C=NC2CCCCC2

InChI

1S/C13H22N2/c1-3-7-12(8-4-1)14-11-15-13-9-5-2-6-10-13/h12-13H,1-10H2

InChI key

QOSSAOTZNIDXMA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Mura et al.
International journal of nanomedicine, 6, 2591-2605 (2011-11-25)
Because of the described hazards related to inhalation of manufactured nanoparticles, we investigated the lung toxicity of biodegradable poly (lactide-co-glycolide) (PLGA) nanoparticles displaying various surface properties on human bronchial Calu-3 cells. Positively and negatively charged as well as neutral nanoparticles
Masashi Toei et al.
The Journal of biological chemistry, 288(36), 25717-25726 (2013-07-31)
N,N-Dicyclohexylcarbodiimide (DCCD) is a classical inhibitor of the F0F1-ATP synthase (F0F1), which covalently binds to the highly conserved carboxylic acid of the proteolipid subunit (c subunit) in F0. Although it is well known that DCCD modification of the c subunit
Denys Pogoryelov et al.
Nature chemical biology, 6(12), 891-899 (2010-10-26)
The microscopic mechanism of coupled c-ring rotation and ion translocation in F(1)F(o)-ATP synthases is unknown. Here we present conclusive evidence supporting the notion that the ability of c-rings to rotate within the F(o) complex derives from the interplay between the
Ryutaro Tokutsu et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(24), 10016-10021 (2013-05-30)
Plants and green algae have a low pH-inducible mechanism in photosystem II (PSII) that dissipates excess light energy, measured as the nonphotochemical quenching of chlorophyll fluorescence (qE). Recently, nonphotochemical quenching 4 (npq4), a mutant strain of the green alga Chlamydomonas
Fatemeh Zeinali Nasrabadi et al.
Molecular diversity, 15(3), 791-798 (2011-03-23)
Reactions of (N-isocyanimino)triphenylphosphorane with N,N-dicyclohexylcarbodiimide in the presence of aromatic (or heteroaromatic) carboxylic acids proceed smoothly at room temperature and in neutral conditions to afford sterically congested 1,3,4-oxadiazole derivatives in high yields. The reaction proceeds smoothly and cleanly under mild

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service