Skip to Content
Merck
All Photos(1)

Key Documents

D2565

Sigma-Aldrich

Diethylenetriaminepentamethylenephosphonic acid heptasodium salt

aqueous solution

Synonym(s):

DTPMPA, [(Phosphonomethyl)imino]bis[2,1-ethanediylnitrilobis(methylene)]tetrakis(phosphonic acid) heptasodium salt

Sign Into View Organizational & Contract Pricing

Select a Size

25 ML
$80.60
500 ML
$122.00

$80.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 ML
$80.60
500 ML
$122.00

About This Item

Empirical Formula (Hill Notation):
C9H21N3O15P5Na7
CAS Number:
Molecular Weight:
727.07
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

$80.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Product Name

Diethylenetriaminepentamethylenephosphonic acid heptasodium salt, 25 % (w/v) in H2O, ~55% (based on active acid and salt), aqueous solution

form

aqueous solution

concentration

25 % (w/v) in H2O
~55% (based on active acid and salt)

color

amber

pH

6.0-8.0 (1% in H2O)

mp

-13 °C

transition temp

freezing point −13 °C

storage temp.

room temp

SMILES string

[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[Na+].[P](=O)(O)(O)CN(C[P](=O)(O)O)CCN(C[P](=O)(O)O)CCN(C[P](=O)(O)O)C[P](=O)(O)O

InChI

1S/C9H28N3O15P5.7Na/c13-28(14,15)5-10(1-3-11(6-29(16,17)18)7-30(19,20)21)2-4-12(8-31(22,23)24)9-32(25,26)27;;;;;;;/h1-9H2,(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)(H2,25,26,27);;;;;;;/q;7*+1

InChI key

OIPXXWBYRWQVLJ-UHFFFAOYSA-N

Application

Used for the removal of metal ions, and may have potential biological properties as a chelator.
Used for the removal of metal ions, and may have potential biological properties as a chelator. Thorium complexes of DTPMPA are preferentially localized in bone rather than soft tissue, offering potential as targeted radiotherapeutic agents.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service