Skip to Content
Merck
All Photos(1)

Key Documents

D2189

Sigma-Aldrich

Recombinant CRM197 expressed in Pseudomonas fluorescens, [Glu52]- Diphtheria toxin

lyophilized powder

Synonym(s):

[Glu52]-Diphtheria toxin from Corynebacterium diphtheriae, CRM 197

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12352202
Pricing and availability is not currently available.

form

lyophilized powder

Quality Level

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Packaging

Package size based on protein content

Physical form

Lyophilized powder containing sodium phosphate, sucrose and polysorbate 80

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Francesco Caiazza et al.
Molecular endocrinology (Baltimore, Md.), 24(5), 953-968 (2010-03-10)
Cytosolic phospholipase A(2)alpha (cPLA(2)alpha) catalyzes the hydrolysis of membrane glycerol-phospholipids to release arachidonic acid as the first step of the eicosanoid signaling pathway. This pathway contributes to proliferation in breast cancer, and numerous studies have demonstrated a crucial role of
Matthias A Oberli et al.
Chemistry & biology, 18(5), 580-588 (2011-05-26)
Nosocomial infections with the Gram-positive pathogen Clostridium difficile pose a major risk for hospitalized patients and result in significant costs to health care systems. Here, we present the chemical synthesis of a PS-II hapten of a cell wall polysaccharide of
Yung-Chih Kuo et al.
Colloids and surfaces. B, Biointerfaces, 91, 242-249 (2011-12-06)
This study investigates the capability of CRM197-grafted polybutylcyanoacrylate (PBCA) nanoparticles (NPs) (CRM197/PBCA NPs) to carry zidovudine (AZT) across the blood-brain barrier (BBB). AZT was loaded on CRM197/PBCA NPs to traverse the monolayer of human brain-microvascular endothelial cells (HBMECs) regulated by
S Rivetti et al.
International journal of immunopathology and pharmacology, 24(3), 639-649 (2011-10-08)
Cross-Reacting Material 197 (CRM197) is a diphtheria toxin non-toxic mutant that has shown antitumor activity in mice and humans. It is still unclear whether this anti-tumorigenic effect depends on its strong inflammatory-immunological property, its ability to inhibit heparin-binding epidermal growth
Mary F Fontana et al.
PLoS pathogens, 12(12), e1006046-e1006046 (2016-12-07)
Dynamic regulation of leukocyte population size and activation state is crucial for an effective immune response. In malaria, Plasmodium parasites elicit robust host expansion of macrophages and monocytes, but the underlying mechanisms remain unclear. Here we show that myeloid expansion

Questions

1–4 of 4 Questions  
  1. How do I reconstitute Product D2189, [Glu52]-Diphtheria toxin from Corynebacterium diphtheriae?

    1 answer
    1. The vial can be reconstituted with 1.0 mL of sterile water. Each vial contains 1 mg of Diphtheria toxin CRM mutant, sodium phosphate buffer, and lactose. It can sometimes be hard to get the product into solution due to the lactose used as a stabilizer. The supplier suggests slight warming (to at least room temperature or 30 °C, but not more than 37 °C) followed by a brief vortex to break up the filaments.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

  3. What is the molecular weight of Product D2189, [Glu52]-Diphtheria toxin from Corynebacterium diphtheriae?

    1 answer
    1. On reducing SDS gels, this protein migrates as a single band of an approximate molecular weight of 63,000 Daltons.

      Helpful?

  4. What kind of mutant, is Product D2189, [Glu52]-Diphtheria toxin from Corynebacterium diphtheriae, and how is this product different from Product No. D0564?

    1 answer
    1. Product No. D0564 is the wild-type toxin and possesses enzymatic activity.  The Glu52 diptheria toxin, also known as CRM197, has a mutation in fragment A (a glycine to glutamic acid substitution at position 52) that results in the complete loss of enzymatic activity. More information on the mutant can be found in the following reference: Uchida, T., Pappenheimer, Jr., A.M. and Harper, A.A.,  J. Biol. Chem., 248, 3851-3854 (1973).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service