Skip to Content
Merck
All Photos(1)

Key Documents

D2043

Sigma-Aldrich

Defensin HNP-1 human

≥80% (HPLC)

Synonym(s):

Neutrophil Peptide-1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C150H222N44O38S6
CAS Number:
Molecular Weight:
3442.03
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥80% (HPLC)

form

powder

composition

Peptide content, ~70%

solubility

water: 1 mg/mL, clear, colorless

UniProt accession no.

antibiotic activity spectrum

fungi
mycobacteria
viruses

Mode of action

cell membrane | interferes

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)[C@@H]2CSSC[C@@H]3NC(=O)[C@H](Cc4ccccc4)NC(=O)[C@H](C)NC(=O)[C@H](Cc5c[nH]c6ccccc56)NC(=O)[C@H](CC(C)C)NC(=O)[C@H](CCCNC(N)=N)NC(=O)CNC(=O)[C@H](CCC(N)=O)NC(=O)[C@H](Cc7ccc(O)cc7)NC(=O)[C@@H](NC(=O)[C@H](CSSC[C@H](NC(=O)[C@H](Cc8ccc(O)cc8)NC(=O)[C@H](CSSC[C@H](NC3=O)C(O)=O)NC(=O)[C@H](C)N)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H]([C@@H](C)CC)C(=O)N9CCC[C@H]9C(=O)N[C@@H](C)C(=O)N2)NC(=O)[C@@H](NC(=O)CNC(=O)[C@H](Cc%10ccc(O)cc%10)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@H](CCC(O)=O)NC(=O)CNC(=O)[C@H](C)NC1=O)[C@@H](C)O)[C@@H](C)CC

InChI

1S/C150H222N44O38S6/c1-14-74(6)116-142(227)171-78(10)121(206)166-64-112(200)173-97(49-51-115(203)204)129(214)175-93(32-23-53-162-148(155)156)127(212)174-94(33-24-54-163-149(157)158)128(213)180-99(59-83-36-42-87(196)43-37-83)125(210)168-66-114(202)190-119(81(13)195)144(229)188-108-71-236-233-68-105-136(221)176-95(34-25-55-164-150(159)160)130(215)193-118(76(8)16-3)145(230)194-56-26-35-110(194)141(226)170-80(12)123(208)185-107(139(224)191-116)70-235-234-69-106(138(223)189-109(146(231)232)72-238-237-67-104(184-120(205)77(9)151)137(222)181-101(135(220)186-105)60-84-38-44-88(197)45-39-84)187-134(219)100(58-82-27-18-17-19-28-82)178-122(207)79(11)169-131(216)103(62-86-63-165-91-30-21-20-29-90(86)91)182-132(217)98(57-73(4)5)179-126(211)92(31-22-52-161-147(153)154)172-113(201)65-167-124(209)96(48-50-111(152)199)177-133(218)102(61-85-40-46-89(198)47-41-85)183-143(228)117(75(7)15-2)192-140(108)225/h17-21,27-30,36-47,63,73-81,92-110,116-119,165,195-198H,14-16,22-26,31-35,48-62,64-72,151H2,1-13H3,(H2,152,199)(H,166,206)(H,167,209)(H,168,210)(H,169,216)(H,170,226)(H,171,227)(H,172,201)(H,173,200)(H,174,212)(H,175,214)(H,176,221)(H,177,218)(H,178,207)(H,179,211)(H,180,213)(H,181,222)(H,182,217)(H,183,228)(H,184,205)(H,185,208)(H,186,220)(H,187,219)(H,188,229)(H,189,223)(H,190,202)(H,191,224)(H,192,225)(H,193,215)(H,203,204)(H,231,232)(H4,153,154,161)(H4,155,156,162)(H4,157,158,163)(H4,159,160,164)/t74-,75-,76-,77-,78-,79-,80-,81+,92-,93-,94-,95-,96-,97-,98-,99-,100-,101-,102-,103-,104-,105-,106-,107-,108-,109-,110-,116-,117-,118-,119-/m0/s1

InChI key

KRJOFJHOZZPBKI-KSWODRSDSA-N

Gene Information

human ... DEFA1(1667)

Amino Acid Sequence

Ala-Cys-Tyr-Cys-Arg-Ile-Pro-Ala-Cys-Ile-Ala-Gly-Glu-Arg-Arg-Tyr-Gly-Thr-Cys-Ile-Tyr-Gln-Gly-Arg-Leu-Trp-Ala-Phe-Cys-Cys [Disulfide bridges: 2−30; 4−19; 9−29]

General description

Chemical structure: peptide
The α-defensin genes, such as DEFA1 (defensin alpha 1) and DEFA3 (defensin α 3) codes for HNP-1, 2 and 3 (human neutrophil peptides).[1] This gene is mapped to human chromosome 8p23.1.[2]

Biochem/physiol Actions

Human neutrophil peptides (HNPs) is involved in endothelial cell dysfunction at the time of early atherosclerotic development. HNP 1−3 can regulate the growth of atherosclerosis. HNP 1−3 levels are usually high in inflammation, including sepsis and acute coronary vascular disorders.[1]
This is an endogenous antibiotic peptide and monocyte chemotactic peptide produced by human neutrophils. Defensins are a family of 3-4 kDa (29-34 amino acids) peptides found in the granules of mammalian phagocytes. The members of this family are variably arginine-rich and share six conserved cysteine residues that participate in intramolecular disulfide bonds.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan J Blower et al.
PLoS neglected tropical diseases, 9(7), e0003862-e0003862 (2015-07-22)
Burkholderia thailandensis is a Gram-negative soil bacterium used as a model organism for B. pseudomallei, the causative agent of melioidosis and an organism classified category B priority pathogen and a Tier 1 select agent for its potential use as a
A J Ouellette
The American journal of physiology, 277(2 Pt 1), G257-G261 (1999-08-13)
The hypothesis that epithelial cells release preformed antibiotic peptides as components of mucosal innate immunity has gained experimental support in recent years. In the mammalian small intestine, Paneth cells secrete granules that are rich in alpha-defensins and additional antimicrobial peptides
Copy number polymorphism and expression level variation of the human alpha-defensin genes DEFA1 and DEFA3
Aldred P M R, et al.
Human Molecular Genetics, 14(14), 2045-2052 (2005)
P S Hiemstra et al.
The European respiratory journal, 12(5), 1200-1208 (1998-12-24)
Neutrophils have the capacity to accumulate in high numbers in the lung during infection and inflammation. Because they play an important role in host defence against infection, but may also cause tissue injury, these cells are thought to be involved
Lingli Tang et al.
Microbes and infection, 17(6), 402-408 (2015-03-11)
Chlamydia trachomatis infection in the lower genital tract, if untreated, can ascend to the upper genital tract, potentially leading to complications such as tubal factor infertility. The ascension involves cell-to-cell spreading, which may require C. trachomatis organisms to overcome mucosal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service