Skip to Content
Merck
All Photos(1)

Key Documents

D2000

Sigma-Aldrich

N-Dansyl-L-serine cyclohexylammonium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H18N2O5S · C6H13N
CAS Number:
Molecular Weight:
437.55
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

form

solid

storage temp.

−20°C

SMILES string

NC1CCCCC1.CN(C)c2cccc3c(cccc23)S(=O)(=O)N[C@@H](CO)C(O)=O

InChI

1S/C15H18N2O5S.C6H13N/c1-17(2)13-7-3-6-11-10(13)5-4-8-14(11)23(21,22)16-12(9-18)15(19)20;7-6-4-2-1-3-5-6/h3-8,12,16,18H,9H2,1-2H3,(H,19,20);6H,1-5,7H2/t12-;/m0./s1

InChI key

NEVMXASXEDTEQO-YDALLXLXSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gaihuan Ren et al.
Journal of colloid and interface science, 540, 177-184 (2019-01-15)
Surfactants with temperature-sensitive polyoxyethylene (POE) chains are widely used to prepare nanoemulsions by the phase inversion temperature (PIT) method. It is therefore anticipated that surfactants with temperature-sensitive polyoxypropylene (POP) chains could also be used to prepare nanoemulsions by the PIT
Job Komen et al.
Lab on a chip, 20(17), 3167-3178 (2020-07-31)
We present a microfluidic device to expose cancer cells to a dynamic, in vivo-like concentration profile of a drug, and quantify efficacy on-chip. About 30% of cancer patients receive drug therapy. In conventional cell culture experiments drug efficacy is tested
Areen M Khattabi et al.
Saudi pharmaceutical journal : SPJ : the official publication of the Saudi Pharmaceutical Society, 26(7), 1022-1026 (2018-11-13)
The objective of this research was to investigate the effect of polymer length on the in vitro characteristics of thymoquinone-melatonin (TQ-MLT) when loaded into our previously prepared targeted drug delivery system (TDDS). Our system constructed from silica nanoparticles (NPs) and
Beata Strachota et al.
Polymers, 11(6) (2019-05-31)
Reversible Diels-Alder (DA) type networks were prepared from furan and maleimide monomers of different structure and functionality. The factors controlling the dynamic network formation and their properties were discussed. Evolution of structure during both dynamic nonequilibrium and isothermal equilibrium network
Georges R Younes et al.
ACS omega, 5(47), 30657-30670 (2020-12-08)
A sol/gel curing method is used in this work to synthesize hybrid partially bio-based polyhydroxyurethanes (PHUs) from dicarbonates derived from glycerol and various diamines. The method consists of end-capping the PHU prepolymers with moisture-sensitive groups, so sealants and adhesives can

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service