Skip to Content
Merck
All Photos(1)

Key Documents

D1947L

Sigma-Aldrich

Diffinity RapidTip®

for PCR Purification, Compatible with Rainin LTS® pipettes

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
41106300

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Diffinity RapidTip effectively removes dNTPs, primers and primer dimers while providing greater than 90% recovery of pure DNA fragments from 100 bp to 10 kb. The functional pipette tip contains everything you need for PCR purification. The tip is filled with a proprietary adsorption technology that has a differential affinity for PCR components. The dispensed solution yields purified, high quality DNA ready for downstream applications such as DNA sequencing, SNP analysis and microarray printing.

Features and Benefits

  • Single step
  • Recovers 90% of high quality dsDNA
  • Optimized for 25 μL RXN

Legal Information

LTS is a registered trademark of Rainin Instrument, LLC
RapidTip is a registered trademark of Diffinity Genomics, Inc.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

STOT RE 1 Inhalation

Target Organs

Lungs

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C N Roterman et al.
Proceedings. Biological sciences, 280(1764), 20130718-20130718 (2013-06-21)
The phylogeny of the superfamily Chirostyloidea (Decapoda: Anomura) has been poorly understood owing to limited taxon sampling and discordance between different genes. We present a nine-gene dataset across 15 chirostyloids, including all known yeti crabs (Kiwaidae), to improve the resolution
Lisa A Santry et al.
Virus research, 175(1), 30-44 (2013-04-16)
Maedi-visna virus (MVV) and caprine arthritis encephalitis virus (CAEV) are related members of a group of small ruminant lentiviruses (SRLVs) that infect sheep and goats. SRLVs are endemic in many countries, including Canada. However, very little is known about the
Marion L East et al.
International journal for parasitology. Parasites and wildlife, 2, 257-265 (2014-02-18)
We provide the first genetic sequence data for a Dipylidium species from a wild carnivore plus an analysis of the effects of ecological, demographic, physiological and behavioural factors on Dipylidium sp. infection prevalence in a social carnivore, the spotted hyaena
Myron T La Duc et al.
Applied and environmental microbiology, 78(16), 5912-5922 (2012-06-26)
Spacecraft hardware and assembly cleanroom surfaces (233 m(2) in total) were sampled, total genomic DNA was extracted, hypervariable regions of the 16S rRNA gene (bacteria and archaea) and ribosomal internal transcribed spacer (ITS) region (fungi) were subjected to 454 tag-encoded
Jason W Sahl et al.
Applied and environmental microbiology, 78(14), 4884-4892 (2012-05-15)
The sequencing and analysis of multiple housekeeping genes has been routinely used to phylogenetically compare closely related bacterial isolates. Recent studies using whole-genome alignment (WGA) and phylogenetics from >100 Escherichia coli genomes has demonstrated that tree topologies from WGA and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service