Skip to Content
Merck
All Photos(1)

Key Documents

D1875

Sigma-Aldrich

Dansyl-L-proline

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20N2O4S
CAS Number:
Molecular Weight:
348.42
Beilstein:
497317
MDL number:
UNSPSC Code:
12352200

storage temp.

−20°C

SMILES string

CN(C)c1cccc2c(cccc12)S(=O)(=O)N3CCC[C@H]3C(O)=O

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Gershkovich et al.
Journal of biochemical and biophysical methods, 45(2), 183-191 (2000-09-16)
A method of conjugating protein molecules under native conditions with water-insoluble hydrophobic compounds is developed. It permits very water-insoluble acids to be gently coupled to the primary amines on proteins or other biopolymers. For this purpose we synthesized a polymer
Alan D Borthwick et al.
Bioorganic & medicinal chemistry letters, 12(13), 1719-1722 (2002-06-18)
Mechanism-based inhibitors of HCMV protease, which are stable to human plasma (> or = 20 h) and have single-figure potency in the microM range against HCMV protease, have been developed based on the dansylproline alpha-methyl pyrrolidine-5,5-trans-lactam nucleus.
Y Abe et al.
Biochimica et biophysica acta, 1433(1-2), 188-197 (1999-08-14)
The enantioselective binding sites on bovine serum albumin were examined by HPLC using 19 racemic 5-N, N-dimethylamino-1-naphthalenesulfonyl derivatives of alpha-amino acids (dansyl amino acids) as chiral probes. On a bovine serum albumin bonded chiral stationary phase, seven L-forms eluted faster
N Okabe et al.
Biological & pharmaceutical bulletin, 17(1), 16-21 (1994-01-01)
Binding properties of Sudlow's site-specific drugs to glycosylated human serum albumin (G-HSA) were investigated using fluorescence and circular dichroism (CD). Dansylamide, phenylbutazone and warfarin were used as site I-specific drugs, and dansylproline, ibuprofen and flufenamic acid were used as site
Yan-Min Chen et al.
Archives of toxicology, 83(3), 255-261 (2008-10-16)
Binding of five perfluoroalkyl acids with human serum albumin (HSA) was investigated by site-specific fluorescence. Intrinsic fluorescence of tryptophan-214 in HSA was monitored upon addition of the chemicals. Although perfluorobutyl acid (PFBA) and perfluorobutane sulfonate (PFBS) did not cause fluorescence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service