Skip to Content
Merck
All Photos(1)

Key Documents

D1542

Sigma-Aldrich

1,4-Dideoxy-1,4-imino-D-arabinitol hydrochloride

enzyme inhibitor

Synonym(s):

2-Hydroxymethyl-3,4-pyrrolidinediol hydrochloride

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
$965.00
25 MG
$2,150.00

$965.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025



Select a Size

Change View
10 MG
$965.00
25 MG
$2,150.00

About This Item

Empirical Formula (Hill Notation):
C5H11NO3 · HCl
CAS Number:
Molecular Weight:
169.61
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

$965.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 01, 2025


Quality Level

Assay

≥98% (TLC)

form

solid

storage condition

under inert gas

solubility

water: 19.60-20.40 mg/mL, clear, colorless to faintly yellow

storage temp.

2-8°C

SMILES string

Cl.OC[C@H]1NC[C@@H](O)[C@@H]1O

InChI

1S/C5H11NO3.ClH/c7-2-3-5(9)4(8)1-6-3;/h3-9H,1-2H2;1H/t3-,4-,5-;/m1./s1

InChI key

PZGVJCJRMKIVLJ-DEVUXVJFSA-N

General description

1,4-Dideoxy-1,4-imino-D-arabinitol is a naturally occurring pyrrolidine alkaloid. It is found in Arachniodes standishii and Angylocalyx boutiqueanus.[1]

Application

1,4-Dideoxy-1,4-imino-D-arabinitol hydrochloride has been used as an α-glucosidase (GAA) inhibitor.[2]

Biochem/physiol Actions

Inhibitor of glycogen phosphorylase and α-glucosidases.

Caution

Extremely hygroscopic, store and handle under argon

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new resorufin-based $\alpha$-glucosidase assay for high-throughput screening
Motabar O, et al.
Analytical Biochemistry, 390(1), 79-84 (2009)
Tetrahedron Letters, 42, 5685-5685 (1986)
The synthesis from d-xylose of the potent and specific enantiomeric glucosidase inhibitors, 1, 4-dideoxy-1, 4-imino-d-arabinitol and 1, 4-dideoxy-1, 4-imin
Fleet GWJ and Smith PW
Tetrahedron, 42(20), 5685-5692 (1986)
Ze Fan et al.
Neuroscience bulletin, 36(12), 1513-1523 (2020-10-14)
General anesthesia severely affects the metabolites in the brain. Glycogen, principally stored in astrocytes and providing the short-term delivery of substrates to neurons, has been implicated as an affected molecule. However, whether glycogen plays a pivotal role in modulating anesthesia-arousal
Junnan Xu et al.
Frontiers in pharmacology, 9, 873-873 (2018-08-23)
Understanding the mechanistic basis for temozolomide (TMZ)-induced glioma resistance is an important obstacle in developing an effective form of chemotherapy for this type of tumor. Glycogenolysis is known to play an essential role in cellular proliferation and potassium homeostasis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service