Skip to Content
Merck
All Photos(1)

Documents

D1129

Sigma-Aldrich

2,3-Dimercapto-1-propanol

Synonym(s):

BAL, British anti-Lewisite, DMP, Dimercaprol, Dithioglycerol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(SH)CH2SH
CAS Number:
Molecular Weight:
124.23
Beilstein:
1732058
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

refractive index

n20/D 1.573 (lit.)

bp

120 °C/15 mmHg (lit.)

density

1.239 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(S)CS

InChI

1S/C3H8OS2/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

WQABCVAJNWAXTE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

233.6 °F - closed cup

Flash Point(C)

112 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raoul Herbrecht et al.
Clinical infectious diseases : an official publication of the Infectious Diseases Society of America, 60(5), 713-720 (2014-11-22)
Strict definition of invasive aspergillosis (IA) cases is required to allow precise conclusions about the efficacy of antifungal therapy. The Global Comparative Aspergillus Study (GCAS) compared voriconazole to amphotericin B (AmB) deoxycholate for the primary therapy of IA. Because predefined
Megan O'Reilly et al.
American journal of physiology. Lung cellular and molecular physiology, 307(12), L948-L958 (2014-10-12)
Bronchopulmonary dysplasia (BPD) is the chronic lung disease of prematurity that affects very preterm infants. Although advances in perinatal care have enabled the survival of infants born as early as 23-24 wk of gestation, the challenge of promoting lung growth
R Vos et al.
American journal of transplantation : official journal of the American Society of Transplantation and the American Society of Transplant Surgeons, 14(12), 2736-2748 (2014-11-15)
Lymphocytic airway inflammation is a major risk factor for chronic lung allograft dysfunction, for which there is no established treatment. We investigated whether azithromycin could control lymphocytic airway inflammation and improve allograft function. Fifteen lung transplant recipients demonstrating acute allograft
Chin-Ming Chen et al.
Journal of thoracic disease, 6(12), 1704-1713 (2015-01-16)
The mortality rate of patients with acute respiratory distress syndrome (ARDS) is still high despite the use of protective ventilatory strategies. We sought to examine the pharmacological effects of glutamine (GLN) in a two-hit model of endotoxin-induced inflammation followed by
James Devaney et al.
Thorax, 70(7), 625-635 (2015-05-20)
Mesenchymal stromal cells (MSCs) demonstrate considerable promise in preclinical acute respiratory distress syndrome models. We wished to determine the efficacy and mechanisms of action of human MSCs (hMSCs) in the setting of acute lung injury induced by prolonged Escherichia coli

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service