Skip to Content
Merck
All Photos(1)

Key Documents

D108

Sigma-Aldrich

1,3-Dipropyl-7-methylxanthine

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18N4O2
CAS Number:
Molecular Weight:
250.30
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

form

solid

color

white

solubility

0.1 M NaOH: soluble
DMSO: soluble

εmax

9,500 at 273 nm

SMILES string

CCCN1C(=O)N(CCC)c2ncn(C)c2C1=O

InChI

1S/C12H18N4O2/c1-4-6-15-10-9(14(3)8-13-10)11(17)16(7-5-2)12(15)18/h8H,4-7H2,1-3H3

InChI key

QVAYTZAGDQIWMB-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Caffeine analog with some selectivity for A2 adenosine receptors.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M T Shamim et al.
Journal of medicinal chemistry, 32(6), 1231-1237 (1989-06-01)
The effects of 8-phenyl and 8-cycloalkyl substituents on the activity of theophylline, caffeine, 1,3-dipropylxanthine, 1,3-dipropyl-7-methylxanthine, 3-propylxanthine, and 1-propylxanthine at A1 adenosine receptors of rat brain and fat cells and at A2 adenosine receptors of rat pheochromocytoma PC12 cells and human
S Hindley et al.
Journal of neuroscience research, 38(4), 399-406 (1994-07-01)
Adenosine and its nucleotides adenosine triphosphate (ATP) and adenosine diphosphate (ADP) stimulate the proliferation of brain astrocytes in vitro and augment the effects of other growth factors. Following brain injury, hypoxia, or around solid tumors with necrotic centers, such as
O Nikodijević et al.
Pharmacology, biochemistry, and behavior, 44(1), 199-216 (1993-01-01)
Chronic ingestion of caffeine by mice caused a marked reduction in locomotor exploratory activity. At least 4 days of withdrawal were required to restore activity to normal levels. Stimulatory effects of injected caffeine were lower in chronically treated mice and
C Blazynski et al.
Journal of neurochemistry, 58(2), 761-767 (1992-02-01)
Much evidence has accumulated supporting the hypothesis that the purine nucleoside adenosine may indeed function as a neuromodulator in the mammalian retina, but to date no reports have directly illustrated a physiological role for this nucleoside. In other regions of
G W Sullivan et al.
Journal of immunology (Baltimore, Md. : 1950), 145(5), 1537-1544 (1990-09-01)
Human rTNF-alpha (greater than or equal to U/ml) decreased PMN nondirected and directed migration to FMLP to approximately 50% of control. Adenosine (100 microM) almost completely restored hrTNF-inhibited migration (nondirected from 54 to 92% and directed migration to from 54

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service