Skip to Content
Merck

D030

R(−)-2,10,11-Trihydroxy-N-propyl-noraporphine hydrobromide hydrate

solid

Synonym(s):

R(−)-2-OH-NPA HBr, R(−)-TNPA HBr

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H21NO3 · HBr · xH2O
CAS Number:
Molecular Weight:
392.29 (anhydrous basis)
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

solid

optical activity

[α]22/D −64°, c = 7 mg/mL in methanol(lit.)

color

light brown

solubility

alcohol: soluble (Solutions should be freshly prepared.), aqueous acid: moderately soluble (Solutions should be freshly prepared), aqueous base: insoluble

storage temp.

2-8°C

SMILES string

Br.[H][C@]12Cc3ccc(O)c(O)c3-c4cc(O)cc(CCN1CCC)c24

Gene Information

human ... DRD2(1813)

Biochem/physiol Actions

Potent, selective D2 dopamine receptor agonist.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



E Chu et al.
Experimental eye research, 69(6), 611-616 (2000-01-06)
The purpose of this study was to correlate potential mechanisms with site(s) of action for TNPA-induced ocular hypotension. In response to R(-)-2, 10, 11-trihydroxy-N-propyl-noraporphine hydrobromide (TNPA, 75 microg), a D2 dopamine receptor agonist, the intraocular pressure decreased by 4.5 and
J L Neumeyer et al.
Journal of medicinal chemistry, 24(7), 898-899 (1981-07-01)
(-)-2,10,11-Trihydroxy-N-n-propylnoraporphine (TNPA,2c) has been synthesized from thebaine (3a), via northebaine (3b), normorphothebaine (2a), and alkylation to the N-propyl derivative 2b. O-Demethylation gave the desired product 2c. Compound 2c showed activity comparable to its 10,11-dihyroxy counterpart (NPA, 1b) on the stimulation
E A Jackson et al.
European journal of pharmacology, 87(1), 15-23 (1983-01-28)
The behavioral actions of some novel aporphines have been examined in rats with selective unilateral 6-hydroxydopamine (6OHDA)-induced destruction of nigrostriatal dopaminergic neurons, and in rats with bilateral 6OHDA-induced destruction of mesolimbic dopaminergic neurons. Dopaminomimetics such as apomorphine (APO) in these